Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03014590_s1
          See other OLFR519 GE Assays ›
          Gene Symbol:
          Olfr519
          Gene Name:
          olfactory receptor 519
          Gene Aliases:
          MOR268-6
          Chromosome Location:
          Chr.7: 108893461 - 108894405 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03014590_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr519
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207160.1 NP_997043.1 1 475 77

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr519

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P525725 NM_207160.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P525725 NM_207160.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P525725 NM_207160.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P525725 NM_207160.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P525725 NM_207160.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P525725 NM_207160.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404951 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404954 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25404928 - -
          Whole Mouse Genome G4122A A_52_P525725 NM_207160.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P525725 NM_207160.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06242029 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06061138 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06061140 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25404960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404951 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404943 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404954 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404921 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06061140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25404962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06242025 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10566708 - -
          Affymetrix Mouse Gene 1.1 ST Array 10566707 - -
          Affymetrix Mouse Clariom S Array PSR0700033565.mm.1 NM_207160.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033565.mm.1 NM_207160.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700033565.mm.1 TC0700004064.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2854265 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline