Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03014882_s1
          See other OLFR524 GE Assays ›
          Gene Symbol:
          Olfr524
          Gene Name:
          olfactory receptor 524
          Gene Aliases:
          MOR103-14P
          Chromosome Location:
          Chr.7: 140201336 - 140205279 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03014882_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr524
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011814.1 NP_001011814.1 1 886 67
          GenBank mRNA BC147558.1 - 1 918 9056549 67
          BC147561.1 - 1 918 9056551 67

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr524

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P438919 NM_001011814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P438919 BC147558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P438919 BC147561.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P438919 NM_001011814.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P438919 BC147558.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P438919 BC147561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P438919 NM_001011814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P438919 BC147558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P438919 BC147561.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P438919 NM_001011814.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P438919 BC147558.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P438919 BC147561.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P438919 NM_001011814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P438919 BC147558.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P438919 BC147561.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P438919 NM_001011814.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P438919 BC147558.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P438919 BC147561.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582856 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582862 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582840 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582844 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582875 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582881 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582871 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582876 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582848 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582867 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582835 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582854 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582860 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582852 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582864 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582834 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582851 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582869 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582831 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582872 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582836 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582838 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582873 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25582886 - -
          Whole Mouse Genome G4122A A_52_P438919 NM_001011814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P438919 BC147558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P438919 BC147561.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P438919 NM_001011814.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P438919 BC147558.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P438919 BC147561.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06179070 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06504291 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06504289 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06504297 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06504298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582843 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582835 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582901 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582838 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25582881 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06504285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05904128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582805 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582890 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582852 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582864 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582807 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658161 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582876 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582818 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582875 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582803 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582872 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05904127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582826 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06368446 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582821 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582884 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582858 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582871 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582886 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06298847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06179058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06631919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582838 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582888 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582810 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582835 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582862 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582854 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06658177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25582814 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06504300 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568955 - -
          Affymetrix Mouse Gene 1.1 ST Array 10568954 - -
          Affymetrix Mouse Clariom S Array PSR0700037804.mm.1 NM_001011814.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037804.mm.1 NM_001011814.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037804.mm.1 TC0700004497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037803.mm.1 TC0700004497.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037805.mm.1 TC0700004497.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160809 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160809 - -
          mouse-6 v1.1 (Illumina) GI_58801393-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160809 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160809 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline