Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03014955_g1
          See other OLFR597 GE Assays ›
          Gene Symbol:
          Olfr597
          Gene Name:
          olfactory receptor 597
          Gene Aliases:
          GA_x5J8B7TU1BB-380-666, MOR23-2
          Chromosome Location:
          Chr.7: 103319139 - 103321431 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03014955_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr597
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011845.2 NP_001011845.2 1 34 70
          GenBank mRNA BC150974.1 - 1 61 9087887 70

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr597

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382856 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382862 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382851 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382859 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25382865 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05956128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382773 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382775 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05956123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382787 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382782 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382768 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382774 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06449604 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25382792 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05956127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05950584 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555784 - -
          Affymetrix Mouse Gene 1.1 ST Array 10555783 - -
          Affymetrix Mouse Clariom S Array PSR0700012744.mm.1 TC0700001489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012743.mm.1 TC0700001489.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012743.mm.1 NM_001011845.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012743.mm.1 NM_001011845.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160543 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160543 - -
          mouse-6 v1.1 (Illumina) GI_58801447-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160543 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160543 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline