Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015018_m1
          See other KCTD14 GE Assays ›
          Gene Symbol:
          Kctd14
          Gene Name:
          potassium channel tetramerisation domain containing 14
          Gene Aliases:
          AI449310, D7Ertd760e
          Chromosome Location:
          Chr.7: 97436380 - 97459557 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015018_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kctd14
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010826.3 NP_001010826.2 2 - 3 349 83
          NM_001012434.3 NP_001012436.1 2 - 3 382 83
          GenBank mRNA AK145279.1 - 2 - 3 350 83
          AY899809.1 - 2 - 3 382 83
          BC024588.1 - 2 - 3 221 5066294 83
          BY735425.1 - 2 - 3 350 83
          CK791017.1 - 2 - 3 270 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700001352.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Kctd14

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964363 CK791017.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964363 AY899809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964363 NM_001012434.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964368 EH106497.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964368 NM_001136235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964368 NM_001010826.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964368 NM_001012434.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964364 NM_001136235.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964364 AY899809.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964364 NM_001010826.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1964364 NM_001012434.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 26159 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 21181 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 491254 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950495 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 470016 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 385105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 77425 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964363 CK791017.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964363 AY899809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964363 NM_001012434.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964364 NM_001136235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964364 AY899809.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964364 NM_001010826.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964364 NM_001012434.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964368 EH106497.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964368 NM_001136235.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964368 NM_001010826.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1964368 NM_001012434.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350537 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350562 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350555 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350565 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350558 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350570 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350560 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350575 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350577 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350540 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350564 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350568 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350552 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350538 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350535 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350571 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350561 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350563 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350548 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350573 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350566 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350539 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350556 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350559 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350557 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25350551 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01583905 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01583914 - -
          Whole Mouse Genome G4122A A_52_P54812 CK791017.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P54812 NM_001136235.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P54812 AY899809.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P54812 NM_001010826.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P54812 NM_001012434.3 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01583905 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06434640 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06226791 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06226797 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06226795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350552 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350515 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350532 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350578 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25350535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06667294 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350527 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05982443 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350540 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350582 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06667292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350565 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06039808 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350538 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350590 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06297849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350562 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350535 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350555 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350585 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350546 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350568 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350558 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350567 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06193427 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350533 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06039806 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06667300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350552 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06667295 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350532 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350557 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350576 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350603 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350525 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350589 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350548 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350573 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350561 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06193425 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350553 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06168223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06587534 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350531 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25350551 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555060 - -
          Affymetrix HT MG-430 PM Array Plate 1455451_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426632_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1426633_PM_s_at - -
          Affymetrix HT MG-430 PM Array Plate 1455452_PM_x_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10555059 - -
          Mouse Genome 430 2.0 Array 1426632_at - -
          Mouse Genome 430 2.0 Array 1426633_PM_s_at - -
          Mouse Genome 430 2.0 Array 1426632_PM_at - -
          Mouse Genome 430 2.0 Array 1426633_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233529_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 233529_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700011460.mm.1 TC0700001352.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011460.mm.1 NM_001010826.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011460.mm.1 NM_001010826.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011460.mm.1 NM_001012434.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011460.mm.1 NM_001012434.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011460.mm.1 NM_001136235.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011460.mm.1 NM_001136235.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011463.mm.1 TC0700001352.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011463.mm.1 NM_001010826.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011463.mm.1 NM_001010826.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011463.mm.1 NM_001012434.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011463.mm.1 NM_001012434.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011463.mm.1 NM_001136235.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011463.mm.1 NM_001136235.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011454.mm.1 TC0700001352.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700011454.mm.1 NM_001010826.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011454.mm.1 NM_001010826.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011454.mm.1 NM_001012434.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700011454.mm.1 NM_001012434.3 Refseq
          Mouse Expression Set 430 1426632_at - -
          Mouse Expression Set 430 1426632_PM_at - -
          Mouse Expression Set 430 1426633_s_at - -
          Mouse Expression Set 430 1426633_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1426633_s_at - -
          Mouse Genome 430A 2.0 Array 1426632_at - -
          Mouse Genome 430A 2.0 Array 1426633_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1426632_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426632_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1426633_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1426632_at - -
          Affymetrix HT Mouse Genome 430A Array 1426633_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1455451_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1455451_at - -
          Affymetrix HT Mouse Genome 430B Array 1455452_PM_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1455452_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 430173 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1223797 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3022911 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3095279 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3095279 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3022911 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline