Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015065_s1
          See other GM5447 GE Assays ›
          Gene Symbol:
          Gm5447
          Gene Name:
          predicted gene 5447
          Gene Aliases:
          EG432743
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015065_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5447
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA AK077314.1 - 1 346 88
          BC147407.1 - 1 305 9056483 88
          BC147408.1 - 1 305 9056476 88

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28920311 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2094277 AK077314.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 836070 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125799 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920333 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920314 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920342 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920336 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920340 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920344 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920349 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920330 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920351 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920345 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920339 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920341 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920332 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920329 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920348 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920337 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920311 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920317 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920326 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28920324 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125799 - -
          Whole Mouse Genome G4122A A_52_P884458 BC147407.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P884458 BC147408.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P884458 AK077314.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P112076 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10646436 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10496080 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10646437 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10646433 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28920311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920353 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920385 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920378 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28920367 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920324 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920311 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10740991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920317 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10697427 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920318 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11011199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920351 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10827312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920312 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920383 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11036973 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920360 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920348 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920339 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920401 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10618938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920325 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920329 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920392 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920321 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920354 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920345 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920369 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920397 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920381 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920365 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11011209 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920330 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920333 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920353 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920374 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920391 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10618943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920364 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920372 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920314 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920385 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920371 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11036972 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920340 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920394 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920387 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920327 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920315 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920313 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920362 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920368 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920389 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920370 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920376 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28920357 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404401 - -
          Affymetrix Mouse Clariom S Array PSR1300002071.mm.1 TC1300000340.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101850364 - -
          Illumina mouse-6 v1.0 expression beadchip 101850364 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224650 - -
          Illumina Sentrix mouse-6.v1 bead array 101850364 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101850364 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2870913 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2870913 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline