Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015239_m1
          See other PMP2 GE Assays ›
          Gene Symbol:
          Pmp2
          Gene Name:
          peripheral myelin protein 2
          Gene Aliases:
          P2
          Chromosome Location:
          Chr.3: 10179851 - 10183885 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015239_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pmp2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001030305.2 NP_001025476.1 3 - 4 412 66
          GenBank mRNA BC099520.1 - 3 - 4 412 30848207 66

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300003223.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pmp2

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          membrane organization
          transporter activity
          fatty acid binding
          lipid binding
          cholesterol binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P89767 BC099520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P89767 NM_001030305.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P89767 BC099520.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P89767 NM_001030305.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 434338 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 296171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 832280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 38132 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P89767 BC099520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P89767 NM_001030305.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P89767 BC099520.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P89767 NM_001030305.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941239 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941218 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941228 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941224 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941221 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941233 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941223 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941208 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941203 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941209 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941206 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941220 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941234 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941248 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21941253 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04824882 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P04824868 - -
          Whole Mouse Genome G4122A A_52_P89767 BC099520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P89767 NM_001030305.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P89767 BC099520.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P89767 NM_001030305.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P04824868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02883746 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02883738 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02883739 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02883747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21941239 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941181 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941236 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941196 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02781297 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02793128 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02626441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02626449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941273 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02508448 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941223 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02881049 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941190 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941197 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02881046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941183 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941200 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941276 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941206 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02779892 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941224 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941192 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02883731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941212 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941252 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02779894 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02883736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941231 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941266 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941199 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941228 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941237 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02781298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02409486 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941189 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941258 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941229 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02508450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941246 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941254 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02508452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941215 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03165203 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941245 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941218 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941216 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03165205 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941198 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941234 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02490351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941259 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02590708 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941272 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02626442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941269 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02883746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941262 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941255 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941179 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02409485 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941210 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02508449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21941202 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10497254 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497256 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497257 - -
          Affymetrix Mouse Gene 1.0 ST Array 10497255 - -
          Affymetrix HT MG-430 PM Array Plate 1458193_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10497253 - -
          Mouse Genome 430 2.0 Array 1458193_PM_at - -
          Mouse Genome 430 2.0 Array 1458193_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18857_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 18857_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0300013020.mm.1 NM_001030305.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013020.mm.1 NM_001030305.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0300013020.mm.1 TC0300003223.mm.1 MTA TC
          Mouse Expression Set 430 1458193_at - -
          Mouse Expression Set 430 1458193_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458193_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1458193_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 476874 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2794342 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2794342 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline