Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03015573_m1
          See other TUBAL3 GE Assays ›
          Gene Symbol:
          Tubal3
          Gene Name:
          tubulin, alpha-like 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 3924635 - 3935277 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03015573_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tubal3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001033879.3 NP_001029051.2 2 - 3 288 81
          GenBank mRNA AK135971.1 - 2 - 3 289 81
          BC147507.1 - 2 - 3 281 9056442 81

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300000015.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Panther Classification:

          Molecular Function -

          tubulin

          Gene Ontology Categories:

          Function(s) Process(es)

          microtubule-based process
          protein polymerization
          nucleotide binding
          GTPase activity
          structural constituent of cytoskeleton
          GTP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049944 CO796832.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049944 NM_001033879.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2049944 AK135971.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 3003 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 842970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 940204 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 456455 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 173020 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 703241 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286605 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29683 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P105671 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049944 CO796832.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049944 NM_001033879.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2049944 AK135971.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790596 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790599 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790586 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790588 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790585 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790593 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790592 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790594 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28790597 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07771472 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P105671 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07771483 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07771483 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P105671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790641 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790567 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790572 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790655 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790581 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790586 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790557 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790601 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790554 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28790704 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790612 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790608 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10448081 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790623 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10987937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790581 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790621 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11035338 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790589 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790614 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10949795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790628 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790611 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790617 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790618 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790586 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790580 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10831366 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790626 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790625 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790599 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790604 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790603 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790598 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790672 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28790574 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10403287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10403290 - -
          Affymetrix Mouse Gene 1.1 ST Array 10403285 - -
          Affymetrix Mouse Clariom S Array PSR1300000106.mm.1 NM_001033879.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000106.mm.1 NM_001033879.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000106.mm.1 TC1300000015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000103.mm.1 NM_001033879.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000103.mm.1 NM_001033879.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000103.mm.1 TC1300000015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000107.mm.1 NM_001033879.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000107.mm.1 NM_001033879.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000107.mm.1 TC1300000015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000104.mm.1 NM_001033879.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000104.mm.1 NM_001033879.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000104.mm.1 TC1300000015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300000105.mm.1 NM_001033879.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000105.mm.1 NM_001033879.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1300000105.mm.1 TC1300000015.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 474982 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244458 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline