| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800009534.mm.1 |
NM_001048217.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009534.mm.1 |
NM_001048217.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009534.mm.1 |
TC1800001279.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800009536.mm.1 |
NM_001048217.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009536.mm.1 |
NM_001048217.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009536.mm.1 |
TC1800001279.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800009535.mm.1 |
NM_001048217.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009535.mm.1 |
NM_001048217.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009535.mm.1 |
TC1800001279.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800009531.mm.1 |
NM_001048217.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009531.mm.1 |
NM_001048217.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009531.mm.1 |
TC1800001279.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800009533.mm.1 |
NM_001048217.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009533.mm.1 |
NM_001048217.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800009533.mm.1 |
TC1800001279.mm.1 |
MTA TC |