Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03016337_m1
          See other HOXD8 GE Assays ›
          Gene Symbol:
          Hoxd8
          Gene Name:
          homeobox D8
          Gene Aliases:
          4921540P06Rik, AI047735, Hox-4.3, Hox-5.4
          Chromosome Location:
          Chr.2: 74704615 - 74707933 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03016337_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hoxd8
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001290731.1 NP_001277660.1 2 - 3 260 66
          NM_008276.3 NP_032302.2 1 - 2 632 66
          GenBank mRNA AK016033.1 - 1 - 2 361 66
          BX513411.1 - 2 - 3 284 66

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the NM_001290731.1 transcripts.

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          multicellular organism development
          anterior/posterior pattern specification
          positive regulation of transcription from RNA polymerase II promoter
          skeletal system morphogenesis
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338953 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21339003 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338954 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21339002 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338949 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338950 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338951 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338952 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977649 NM_008276.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977649 AK015019.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1977649 AK016033.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 124221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32213 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155793 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166843 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P117979 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977649 NM_008276.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977649 AK015019.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1977649 AK016033.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P485542 NM_008276.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P485542 AK015019.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P485542 AK016033.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485542 NM_008276.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485542 AK015019.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P485542 AK016033.1 mouse_transcriptome
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338949 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338953 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338950 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338954 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21339003 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338955 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21339002 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338956 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338952 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21338951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338893 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338959 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338923 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338963 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338877 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338988 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338961 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338888 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339031 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338905 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21339043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338957 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21338913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338943 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338935 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338959 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338956 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338957 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338913 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338936 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338931 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01892942 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338945 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02096764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338921 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338919 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338963 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338928 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338916 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338940 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338922 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338923 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338966 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01827637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338926 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338915 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338914 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21338918 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10472969 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472966 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472968 - -
          Affymetrix Mouse Gene 1.0 ST Array 10472967 - -
          Affymetrix HT MG-430 PM Array Plate 1431099_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1433638_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10472965 - -
          Mouse Genome 430 2.0 Array 1431099_at - -
          Mouse Genome 430 2.0 Array 1433638_PM_s_at - -
          Mouse Genome 430 2.0 Array 1433638_s_at - -
          Mouse Genome 430 2.0 Array 1431099_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15437_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 15437_at - -
          Affymetrix Mouse Clariom S Array PSR0200008859.mm.1 NM_008276.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008859.mm.1 TC0200001096.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008855.mm.1 NM_008276.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008855.mm.1 TC0200001096.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008857.mm.1 NM_008276.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008857.mm.1 TC0200001096.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008860.mm.1 NM_008276.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008860.mm.1 TC0200001096.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200008856.mm.1 NM_008276.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0200008856.mm.1 TC0200001096.mm.1 MTA TC
          Mouse Expression Set 430 1431099_at - -
          Mouse Expression Set 430 1431099_PM_at - -
          Mouse Expression Set 430 1433638_PM_s_at - -
          Mouse Expression Set 430 1433638_s_at - -
          Murine Genome U74v2 Set 93221_at - -
          Murine Genome U74v2 Set 93221_PM_at - -
          Mouse Genome 430A 2.0 Array 1431099_PM_at - -
          Mouse Genome 430A 2.0 Array 1431099_at - -
          Affymetrix HT Mouse Genome 430A Array 1431099_at - -
          Affymetrix HT Mouse Genome 430A Array 1431099_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1433638_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1433638_PM_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 567608 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2693052 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2693052 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2693052 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline