Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03017045_s1
          See other OLFR44 GE Assays ›
          Gene Symbol:
          Olfr44
          Gene Name:
          olfactory receptor 44
          Gene Aliases:
          IB3, MOR171-5
          Chromosome Location:
          Chr.9: 39484087 - 39493988 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03017045_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr44
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146830.2 NP_667041.2 2 447 144
          GenBank mRNA AF102518.1 - 1 226 144
          CB174038.1 - 2 454 144

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr44

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014395 NM_146830.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014395 AF102518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2014395 CB173511.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 75849 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 430459 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 297259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 926222 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 611953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 104419 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 20800 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P123856 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014395 NM_146830.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014395 AF102518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2014395 CB173511.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P257690 NM_146830.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P257690 AF102518.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P257690 CB174038.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438716 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438721 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438725 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438719 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438727 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438735 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438710 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438738 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438728 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438706 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438713 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438717 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438702 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438712 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438694 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438709 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438696 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438724 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438733 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438739 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438732 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438723 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438730 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26438708 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P123856 - -
          Whole Mouse Genome G4122A A_51_P257690 NM_146830.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257690 AF102518.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P257690 CB174038.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P116917 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P115300 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P123856 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07581122 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07581120 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156847 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08156840 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07581126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438708 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26438710 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438758 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438752 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438741 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438745 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438730 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438747 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438732 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438739 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438760 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438764 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438746 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438717 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438723 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07657228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438724 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438749 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438763 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438761 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438744 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438742 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438755 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438735 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438754 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438765 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438751 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438738 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438757 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26438728 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10592411 - -
          Affymetrix Mouse Gene 1.1 ST Array 10592410 - -
          Affymetrix Mouse Clariom S Array PSR0900017203.mm.1 NM_146830.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017203.mm.1 NM_146830.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017203.mm.1 TC0900002078.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0900017204.mm.1 NM_146830.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017204.mm.1 NM_146830.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0900017204.mm.1 TC0900002078.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5890685 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1222244 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1222244 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1222244 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-4lm45:80/100.66.75.98:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0