Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03019165_s1
          See other SBPL GE Assays ›
          Gene Symbol:
          Sbpl
          Gene Name:
          spermine binding protein-like
          Gene Aliases:
          2310068J22Rik
          Chromosome Location:
          Chr.17: 23953085 - 23955273 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03019165_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sbpl
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001077421.1 NP_001070889.1 3 557 124
          GenBank mRNA AK009270.1 - 3 558 123
          AK009356.1 - 3 547 124
          AK009376.1 - 3 558 124
          AK009398.1 - 3 559 124
          AK009406.1 - 3 558 124
          AK009446.1 - 3 558 124
          AK009467.1 - 3 558 124
          AK009489.1 - 3 558 124
          AK009501.1 - 3 556 124
          AK009509.1 - 3 558 124
          AK009539.1 - 3 558 124
          AK009553.1 - 3 557 124
          AK009598.1 - 3 558 124
          AK009609.1 - 3 558 124
          AK009613.1 - 3 558 124
          AK009627.1 - 3 560 124
          AK009630.1 - 3 557 124
          AK009631.1 - 3 561 124
          AK009647.1 - 3 556 124
          AK009712.1 - 3 558 124
          AK009758.1 - 3 556 124
          AK009761.1 - 3 556 124
          AK009819.1 - 3 124
          AK009834.1 - 3 558 124
          AK009845.1 - 3 558 124
          AK009910.1 - 3 557 124
          AK009917.1 - 3 560 125
          AK009931.1 - 3 560 124
          AK009982.1 - 3 557 124
          AK010076.1 - 3 557 124
          AK010078.1 - 3 558 124
          AK010083.1 - 3 558 124
          AK010102.1 - 3 558 124
          AK010114.1 - 3 558 124
          AK010161.1 - 3 558 124
          AK010189.1 - 3 558 124
          AK010216.1 - 3 558 124
          AK010221.1 - 3 557 124
          AK010237.1 - 3 557 124
          AK075878.1 - 2 437 124
          AK075905.1 - 1 286 124
          AK160518.1 - 3 557 124
          BC089471.1 - 3 555 30301294 124

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003092 AK160518.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003092 BC089471.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003092 AK010189.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003092 AK010216.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2003092 AK010161.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779820 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 934490 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P161358 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003092 AK160518.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003092 BC089471.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003092 AK010189.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003092 AK010216.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2003092 AK010161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169795 BC089471.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169795 AK010189.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169795 AK010216.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169795 AK010161.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P169795 AK010221.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169795 BC089471.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169795 AK010189.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169795 AK010216.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169795 AK010161.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P169795 AK010221.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P126936 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13385167 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13385144 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13385150 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13385159 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13385164 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13385140 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13385152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094513 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094602 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094550 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094591 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094571 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094576 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094574 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094564 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094583 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094585 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31094516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094531 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13137093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13523089 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094576 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094551 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094579 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13477912 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13444053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13477919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094568 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094558 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094587 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094563 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13385144 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094516 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094553 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094557 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094574 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094564 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13477926 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094571 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094547 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094594 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094549 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094577 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094572 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094592 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13586244 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31094588 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10448401 - -
          Affymetrix Mouse Gene 1.0 ST Array 10448400 - -
          Affymetrix Mouse Gene 1.1 ST Array 10448399 - -
          Affymetrix Mouse Clariom S Array PSR1700014114.mm.1 NM_001077421.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014114.mm.1 NM_001077421.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014114.mm.1 TC1700001610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014115.mm.1 NM_001077421.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014115.mm.1 NM_001077421.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014115.mm.1 TC1700001610.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700014113.mm.1 NM_001077421.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014113.mm.1 NM_001077421.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700014113.mm.1 TC1700001610.mm.1 MTA TC
          Mouse Expression Set 430 1418307_PM_a_at - -
          Mouse Expression Set 430 1418307_a_at - -
          Mouse Genome 430A 2.0 Array 1418307_a_at - -
          Mouse Genome 430A 2.0 Array 1418307_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 745943 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 107050278 - -
          Illumina mouse-6 v1.0 expression beadchip 107050278 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1231356 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1231356 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2549559 - -
          Illumina Sentrix mouse-6.v1 bead array 107050278 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 107050278 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1253051 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline