Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03019541_u1
          See other AKR1C12 GE Assays ›
          Gene Symbol:
          Akr1c12
          Gene Name:
          aldo-keto reductase family 1, member C12
          Gene Aliases:
          AI266976, Akra
          Chromosome Location:
          Chr.13: 4268172 - 4279399 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03019541_u1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Akr1c12
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013777.2 NP_038805.2 9 1055 146
          GenBank mRNA AK007716.1 - 9 1053 146
          BC012643.1 - 9 1029 4205715 146
          BC063780.1 - 9 1032 4219625 146

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1300001378.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Amplicon spans exons and probe does not span exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Akr1c12

          Panther Classification:

          Molecular Function -

          oxidoreductase reductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          xenobiotic metabolic process
          oxidation-reduction process
          aldo-keto reductase (NADP) activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 AB027237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 AF177041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 BC012643.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 BC063780.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 AB027237.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 AF177041.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 BC012643.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P242859 BC063780.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477682 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 637813 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 944500 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 511796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806257 AK035245.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806257 AK050961.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 140862 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P144865 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P152331 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 AB027237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 AF177041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 BC012643.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 BC063780.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 AB027237.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 AF177041.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 BC012643.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P242859 BC063780.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792009 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792024 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792008 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792023 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792019 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792025 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792017 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792010 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792028 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792020 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28792005 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P144865 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P152331 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07772101 - -
          Whole Mouse Genome G4122A A_51_P242859 AB027237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242859 AF177041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242859 BC012643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242859 BC063780.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242859 AB027237.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242859 AF177041.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242859 BC012643.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P242859 BC063780.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P152331 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P144865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10981669 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10981659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791954 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791948 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791906 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791898 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28792009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28791981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792031 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792023 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791962 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791964 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791971 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792002 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10885597 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792019 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791966 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791975 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791981 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792021 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792020 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10766913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791978 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791958 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792008 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792013 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791980 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791991 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791986 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792007 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791969 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792028 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11006715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791974 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792017 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28792009 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10700168 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28791984 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10407451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407446 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407452 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407449 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407448 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407450 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10407447 - -
          Affymetrix HT MG-430 PM Array Plate 1422000_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10407445 - -
          Mouse Genome 430 2.0 Array 1422000_PM_at - -
          Mouse Genome 430 2.0 Array 1450455_PM_s_at - -
          Mouse Genome 430 2.0 Array 1450455_s_at - -
          Mouse Genome 430 2.0 Array 1422000_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 622402_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 622402_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300008645.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008645.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008645.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008647.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008647.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008647.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008641.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008641.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008641.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008644.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008644.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008644.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008643.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008643.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008643.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008648.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008648.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008648.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008642.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008642.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008642.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008646.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008646.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008646.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008640.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008640.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008640.mm.1 TC1300001378.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300008650.mm.1 NM_013777.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008650.mm.1 NM_013777.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300008650.mm.1 TC1300001378.mm.1 MTA TC
          Mouse Expression Set 430 1422000_at - -
          Mouse Expression Set 430 1450455_s_at - -
          Mouse Expression Set 430 1422000_PM_at - -
          Mouse Expression Set 430 1450455_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1422000_at - -
          Mouse Genome 430A 2.0 Array 1450455_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1422000_PM_at - -
          Mouse Genome 430A 2.0 Array 1450455_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1422000_at - -
          Affymetrix HT Mouse Genome 430A Array 1422000_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 586968 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 6520593 - -
          Illumina mouse-6 v1.0 expression beadchip 540068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000540068 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006520593 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1254015 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1254015 - -
          mouse-6 v1.1 (Illumina) scl44313.2.1_9-S - -
          mouse-6 v1.1 (Illumina) scl0027383.2_307-S - -
          Illumina Sentrix mouse-6.v1 bead array 540068 - -
          Illumina Sentrix mouse-6.v1 bead array 6520593 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0027383.2_307-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44313.2.1_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 540068 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6520593 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1254015 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2606079 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1254015 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2606079 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0