| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100023646.mm.1 |
NM_008381.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023646.mm.1 |
TC0100002945.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023647.mm.1 |
NM_008381.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023647.mm.1 |
TC0100002945.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023649.mm.1 |
NM_008381.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023649.mm.1 |
TC0100002945.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100023648.mm.1 |
NM_008381.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100023648.mm.1 |
TC0100002945.mm.1 |
MTA TC |