Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03024120_m1
          See other DAND5 GE Assays ›
          Gene Symbol:
          Dand5
          Gene Name:
          DAN domain family, member 5
          Gene Aliases:
          Cerl-2, Cerr2, Dte, coco
          Chromosome Location:
          Chr.8: 84815405 - 84832265 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03024120_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dand5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_201227.3 NP_957679.1 1 - 2 455 157
          GenBank mRNA AB070694.1 - 1 - 2 312 157
          AY387409.1 - 1 - 2 451 157
          BC115659.1 - 1 - 2 404 40103255 157
          BC115660.2 - 1 - 2 157

          Back To Top

          More Information


          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dand5

          Gene Ontology Categories:

          Function(s) Process(es)

          determination of left/right asymmetry in lateral mesoderm
          ventricular septum development
          atrial septum development
          determination of left/right symmetry
          regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of transforming growth factor beta receptor signaling pathway
          negative regulation of BMP signaling pathway
          sequestering of nodal from receptor via nodal binding
          determination of heart left/right asymmetry
          negative regulation of nodal signaling pathway
          nodal signaling pathway involved in determination of lateral mesoderm left/right asymmetry
          protein binding
          morphogen activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022160 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017526 NM_201227.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017526 NR_033145.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017526 AK143395.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2017526 AY387409.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 432119 AK089636.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 950708 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 57326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742610 NM_001081194.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 388544 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179455 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P159397 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017526 NM_201227.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017526 NR_033145.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017526 AK143395.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2017526 AY387409.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P120140 BC115659.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P120140 BC115660.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P120140 AB070694.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P120140 AY387409.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022130 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022116 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022141 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022133 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022121 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022131 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022143 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022119 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022136 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022148 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022138 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022127 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022139 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022123 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022142 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022125 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022122 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022118 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022129 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022134 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26022153 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P159397 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06579278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06579284 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01784190 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179455 - -
          Whole Mouse Genome G4122A A_52_P120140 BC115659.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P120140 BC115660.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P120140 AB070694.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P120140 AY387409.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179455 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P159397 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P06579278 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07128370 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07319799 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07256250 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06809815 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07319796 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07319793 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07042969 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06830264 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26022160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022102 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022118 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022100 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022085 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022124 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022133 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022101 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022114 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022130 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26022072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022146 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022110 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07042969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07319796 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07252033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06984282 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022089 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07042968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022142 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022095 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022130 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06734947 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07042971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06772529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07413630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07133276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022083 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022103 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022143 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022090 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022121 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022124 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06734952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022071 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022118 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022108 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06946819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26022102 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06734950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07253734 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10580206 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580205 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580204 - -
          Affymetrix Mouse Gene 1.0 ST Array 10580207 - -
          Affymetrix HT MG-430 PM Array Plate 1436476_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10580202 - -
          Mouse Genome 430 2.0 Array 1436476_PM_at - -
          Mouse Genome 430 2.0 Array 1436476_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23863_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 23863_PM_at - -
          Mouse Expression Set 430 1436476_at - -
          Mouse Expression Set 430 1436476_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436476_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1436476_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 888138 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2850072 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002850072 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2754305 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2754305 - -
          mouse-6 v1.1 (Illumina) scl023863.2_102-S - -
          Illumina Sentrix mouse-6.v1 bead array 2850072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl023863.2_102-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2850072 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2754305 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2754305 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-9kpsn:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0