Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03024883_sH
          See other TAAR7B GE Assays ›
          Gene Symbol:
          Taar7b
          Gene Name:
          trace amine-associated receptor 7B
          Gene Aliases:
          Gm698, taR-7b
          Chromosome Location:
          Chr.10: 23999939 - 24001015 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03024883_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Taar7b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001010827.1 NP_001010827.1 1 542 60

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Taar7b

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          G-protein coupled receptor signaling pathway
          sensory perception of chemical stimulus
          positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway
          trace-amine receptor activity
          signal transducer activity
          G-protein coupled receptor activity
          G-protein coupled amine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2118655 NM_001010827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2118655 NM_001010827.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27006308 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08366162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006266 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27006247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006288 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006280 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006308 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006305 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08562737 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006297 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08574281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08366162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006309 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27006303 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10368282 - -
          Affymetrix Mouse Gene 1.0 ST Array 10362159 - -
          Affymetrix Mouse Gene 1.1 ST Array 10362158 - -
          Affymetrix Mouse Gene 1.1 ST Array 10368281 - -
          Affymetrix Mouse Clariom S Array PSR1000001452.mm.1 NM_001010827.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001452.mm.1 NM_001010827.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000001452.mm.1 TC1000000199.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 895028 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2926408 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2926408 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-h9fps:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline