Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03029652_s1
          See other MAPK1IP1 GE Assays ›
          Gene Symbol:
          Mapk1ip1
          Gene Name:
          mitogen-activated protein kinase 1 interacting protein 1
          Gene Aliases:
          2310009E07Rik, AU043776, MISS
          Chromosome Location:
          Chr.7: 138835818 - 138846267 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03029652_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mapk1ip1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001045483.1 NP_001038948.1 2 1149 92
          NM_027115.2 NP_081391.1 2 1045 92
          GenBank mRNA AK009250.1 - 2 1036 92
          BC048375.1 - 2 1142 92
          BC055699.1 - 3 1513 92
          BC069850.1 - 1 713 5366053 92

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mapk1ip1

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          biological_process
          meiotic cell cycle
          identical protein binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574331 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574421 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574420 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574325 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574419 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574417 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2142151 AK009250.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2142151 AW047538.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2142151 BC048375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2142151 BC055699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2142151 BC069850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119716 AW047538.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119716 BC048375.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119716 BC055699.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119716 BC069850.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2119716 NM_001045483.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 964259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404822 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150953 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 742194 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 72454 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 484071 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 179031 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 102623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 151737 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P128790 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138362 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2142151 AK009250.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2142151 AW047538.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2142151 BC048375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2142151 BC055699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2142151 BC069850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119716 AW047538.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119716 BC048375.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119716 BC055699.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119716 BC069850.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2119716 NM_001045483.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574410 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574401 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574421 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574412 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574405 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574403 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574402 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574417 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574395 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574413 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574416 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574414 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574415 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574390 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574398 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574407 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574419 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574411 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574389 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25574404 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P128790 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138362 - -
          Whole Mouse Genome G4122A A_51_P467682 AK009250.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467682 AW047538.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467682 BC048375.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467682 BC055699.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P467682 BC069850.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104396 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138362 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P128790 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06328627 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06328634 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06491784 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06328626 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574420 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574419 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574417 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574421 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574325 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25574331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574375 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574421 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574347 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574398 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574404 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574356 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25574417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06328626 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574404 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574415 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574398 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05990250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06508934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06309129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06309134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06508937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574418 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574395 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574383 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574314 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574325 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574414 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06278215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574312 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574413 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06508940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574317 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574417 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574410 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574412 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574334 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574389 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06508928 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25574343 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10568782 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568783 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568784 - -
          Affymetrix Mouse Gene 1.0 ST Array 10568781 - -
          Affymetrix HT MG-430 PM Array Plate 1452798_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10568780 - -
          Mouse Genome 430 2.0 Array 1452798_PM_s_at - -
          Mouse Genome 430 2.0 Array 1452798_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69546_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 69546_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700037502.mm.1 NM_001045483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037502.mm.1 NM_001045483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037502.mm.1 NM_027115.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037502.mm.1 NM_027115.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037502.mm.1 TC0700004473.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700037505.mm.1 NM_001045483.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037505.mm.1 NM_001045483.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700037505.mm.1 TC0700004473.mm.1 MTA TC
          Mouse Expression Set 430 1452798_s_at - -
          Mouse Expression Set 430 1452798_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1452798_PM_s_at - -
          Affymetrix HT Mouse Genome 430B Array 1452798_s_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 460372 AK002243.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 460372 AK010305.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 460372 BC011131.1 genbank_mrna_mouse
          Illumina mouse-6 v1.0 expression beadchip 460372 NM_025886.3 refseq_mrna_mouse_NM
          Illumina mouseRef-8 v1.0 expression beadchip 0000460372 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1224936 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1224936 - -
          mouse-6 v1.1 (Illumina) scl30536.3_403-S - -
          Illumina Sentrix mouse-6.v1 bead array 460372 AK002243.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 460372 AK010305.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 460372 BC011131.1 genbank_mrna_mouse
          Illumina Sentrix mouse-6.v1 bead array 460372 NM_025886.3 refseq_mrna_mouse_NM
          Illumina Sentrix MouseRef-8 Expression BeadChip scl30536.3_403-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460372 AK002243.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460372 AK010305.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460372 BC011131.1 genbank_mrna_mouse
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 460372 NM_025886.3 refseq_mrna_mouse_NM
          MouseRef-8 v2 Expression BeadChip ILMN_1224936 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3064669 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3142573 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3064669 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1224936 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3142573 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl30536.3_403-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline