Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03031040_s1
          See other TMEM174 GE Assays ›
          Gene Symbol:
          Tmem174
          Gene Name:
          transmembrane protein 174
          Gene Aliases:
          0610009B10Rik, AA986094
          Chromosome Location:
          Chr.13: 98634978 - 98637410 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03031040_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem174
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_026685.2 NP_080961.1 1 620 96
          GenBank mRNA AK002365.1 - 1 547 96
          AK057630.1 - 1 548 96
          BC030878.1 - 1 569 4241631 96

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem174

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 AK057630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 BC030878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 NM_026685.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 AK002365.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 AK057630.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 BC030878.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 NM_026685.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P492346 AK002365.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540135 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 753968 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 423283 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P172421 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 AK057630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 BC030878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 NM_026685.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 AK002365.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 AK057630.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 BC030878.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 NM_026685.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P492346 AK002365.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258754 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258756 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258782 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258757 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258774 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258785 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258778 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258766 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258750 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258772 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258779 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258770 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258788 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258752 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258787 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258755 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29258776 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P172421 - -
          Whole Mouse Genome G4122A A_51_P492346 AK057630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492346 BC030878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492346 NM_026685.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492346 AK002365.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492346 AK057630.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492346 BC030878.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492346 NM_026685.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P492346 AK002365.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P172421 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10675642 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10675647 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10582094 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10714749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258757 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258709 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29258720 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258713 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258731 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11083585 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258734 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258750 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258745 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258703 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258774 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258710 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258740 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258747 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258776 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11012279 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258758 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258782 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258787 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258761 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10641422 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258757 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258754 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258752 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258707 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258792 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258766 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258779 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258785 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258759 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258730 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258769 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258728 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10867323 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258794 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258733 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258712 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10675647 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258778 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10582097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258795 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258791 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258763 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258797 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258780 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258756 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258715 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258799 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258788 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258739 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258790 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258709 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10641423 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258737 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258786 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258803 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10781483 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29258755 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10411458 - -
          Affymetrix Mouse Gene 1.0 ST Array 10411457 - -
          Affymetrix HT MG-430 PM Array Plate 1435201_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1453031_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10411456 - -
          Mouse Genome 430 2.0 Array 1435201_at - -
          Mouse Genome 430 2.0 Array 1453031_PM_at - -
          Mouse Genome 430 2.0 Array 1453031_at - -
          Mouse Genome 430 2.0 Array 1435201_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68344_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68344_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1300016820.mm.1 NM_026685.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016820.mm.1 NM_026685.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016820.mm.1 TC1300002505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300016821.mm.1 NM_026685.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016821.mm.1 NM_026685.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016821.mm.1 TC1300002505.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1300016819.mm.1 NM_026685.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016819.mm.1 NM_026685.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1300016819.mm.1 TC1300002505.mm.1 MTA TC
          Mouse Expression Set 430 1435201_at - -
          Mouse Expression Set 430 1453031_at - -
          Mouse Expression Set 430 1453031_PM_at - -
          Mouse Expression Set 430 1435201_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435201_at - -
          Affymetrix HT Mouse Genome 430B Array 1453031_at - -
          Affymetrix HT Mouse Genome 430B Array 1453031_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1435201_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 545165 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1216425 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216425 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline