| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30485071 |
- |
- |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018101 |
BC047149.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018101 |
BC055034.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018101 |
BC060144.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018101 |
AK076169.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2018101 |
BC033446.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2172822 |
AK008925.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
812226 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK018533.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK036488.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK146004.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK150370.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK152477.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK153166.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK157249.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
AK159835.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
BC005450.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
M32489.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
636566 |
NM_008320.3 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
18924 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018101 |
BC047149.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018101 |
BC055034.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018101 |
BC060144.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018101 |
AK076169.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2018101 |
BC033446.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485069 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485042 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485071 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485036 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485058 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485037 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485079 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485045 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485068 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485048 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485066 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485049 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485039 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485052 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485060 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485035 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485056 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P30485062 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03114798 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P08481192 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03114818 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P03114808 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P205695 |
BC047149.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P205695 |
BC055034.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P205695 |
BC060144.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P205695 |
AK076169.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P205695 |
BC033446.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P218137 |
AK008925.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P359301 |
AK087309.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P136010 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P03114798 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P03114818 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12319152 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11959776 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12248216 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P11959779 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12319155 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P12319158 |
- |
- |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P30485071 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485035 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485148 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485048 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485045 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485062 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485058 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485037 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485071 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485137 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485161 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485028 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485060 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485047 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485151 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485056 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485141 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485152 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485068 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485166 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485144 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485128 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485155 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485156 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485099 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485110 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485049 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485092 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485069 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485154 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485039 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485066 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P30485143 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485071 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485039 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11927706 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485092 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485056 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485069 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12359754 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11917332 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485098 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485079 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12420898 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12132519 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485030 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485042 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485047 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11988133 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P11959696 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12053048 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485052 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485028 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485043 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485068 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485045 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485062 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12053049 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485034 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12079100 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12248213 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485027 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485035 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P12359751 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485060 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485076 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485033 |
- |
- |
| Agilent Six_ChIP_03_of_03_v2 |
A_68_P30485066 |
- |
- |