Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03031209_s1
          See other 9130008F23RIK GE Assays ›
          Gene Symbol:
          9130008F23Rik
          Gene Name:
          RIKEN cDNA 9130008F23 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 40875482 - 40880558 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03031209_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: 9130008F23Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027834.3 NP_082110.1 1 466 80
          GenBank mRNA AK018601.1 - 1 467 80
          AK033670.1 - 1 462 80
          BC132063.1 - 1 462 40130340 80
          BC132065.1 - 1 462 40130342 80

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene 9130008F23Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177906 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177907 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499028 NM_027834.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499028 AK018601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499028 AK033670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499028 NM_027834.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499028 AK018601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P499028 AK033670.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 38407 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 635872 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255807 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42598 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 176912 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P107115 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499028 NM_027834.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499028 AK018601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499028 AK033670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499028 NM_027834.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499028 AK018601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P499028 AK033670.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177907 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177920 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177901 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177896 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177899 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177910 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177900 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177912 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177919 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177924 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177916 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177922 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177918 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177915 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177897 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177933 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177902 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177931 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177898 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177909 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177926 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177921 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177929 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177935 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177904 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177937 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P31177936 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P107115 - -
          Whole Mouse Genome G4122A A_52_P499028 NM_027834.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499028 AK018601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499028 AK033670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499028 NM_027834.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499028 AK018601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P499028 AK033670.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P104916 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P107115 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13199867 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13199859 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13404204 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13199865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13404197 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P13199864 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177904 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177906 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177905 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P31177907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13492916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13490145 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177812 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177887 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177945 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177883 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13314136 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177932 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177874 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177925 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177942 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177885 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177901 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177939 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177826 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177824 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177898 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177869 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177935 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177832 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177814 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177916 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177922 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177947 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177890 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177892 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177915 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13162044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13170954 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177900 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177868 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13199865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177940 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177870 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177929 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13170956 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177865 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177876 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177948 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177907 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177919 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177863 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177899 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177896 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177904 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177918 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13361743 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177938 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177837 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13211673 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13199864 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177936 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177897 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177913 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177902 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177949 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177829 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177833 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177934 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177937 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13404197 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177920 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177880 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13474378 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177906 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177884 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177943 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31177926 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10450949 - -
          Affymetrix Mouse Gene 1.0 ST Array 10450950 - -
          Affymetrix HT MG-430 PM Array Plate 1430221_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10450948 - -
          Mouse Genome 430 2.0 Array 1430221_at - -
          Mouse Genome 430 2.0 Array 1430221_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71583_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 71583_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1700019746.mm.1 NM_027834.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019746.mm.1 NM_027834.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019746.mm.1 TC1700002107.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019747.mm.1 NM_027834.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019747.mm.1 NM_027834.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019747.mm.1 TC1700002107.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700019748.mm.1 NM_027834.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019748.mm.1 NM_027834.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1700019748.mm.1 TC1700002107.mm.1 MTA TC
          Mouse Expression Set 430 1430221_at - -
          Mouse Expression Set 430 1430221_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430221_at - -
          Affymetrix HT Mouse Genome 430B Array 1430221_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 606186 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102030537 - -
          Illumina mouse-6 v1.0 expression beadchip 102030537 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1236953 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220766 - -
          Illumina Sentrix mouse-6.v1 bead array 102030537 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102030537 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2867735 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220766 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1236953 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2867735 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline