Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03031360_s1
          See other SAMT3 GE Assays ›
          Gene Symbol:
          Samt3
          Gene Name:
          spermatogenesis associated multipass transmembrane protein 3
          Gene Aliases:
          1700072E05Rik
          Chromosome Location:
          Chr.X: 86044199 - 86047313 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03031360_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Samt3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028554.3 NP_082830.2 3 847 113
          GenBank mRNA AK006935.1 - 3 848 113

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Samt3

          Panther Classification:

          Molecular Function -

          tight junction

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060553 NM_028554.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060553 AK006935.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060552 NM_028554.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2060552 AK006935.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 668881 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 109398 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P109427 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060553 NM_028554.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060553 AK006935.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060552 NM_028554.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2060552 AK006935.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508180 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508217 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508202 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508194 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508204 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508212 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508214 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508200 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508199 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508192 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508211 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508191 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508196 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508190 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508216 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508183 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508185 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508188 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508207 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508210 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508215 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508193 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508195 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508187 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32508174 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P109427 - -
          Whole Mouse Genome G4122A A_51_P306565 NM_028554.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P306565 AK006935.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106273 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P109427 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14522868 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14522870 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14522865 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15195932 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14522866 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508165 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508207 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508180 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508202 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508248 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508158 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508155 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32508222 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508157 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508231 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508183 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508211 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508228 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508216 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508203 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508160 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508188 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508181 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508224 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508235 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508227 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508219 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508215 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508217 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508208 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14763352 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508191 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508223 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508153 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508200 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14532112 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508192 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14763343 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508180 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508233 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508167 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508204 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508236 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508171 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508178 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508161 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508214 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508175 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508158 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508182 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508193 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508196 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15178086 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508190 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508165 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508199 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14763342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14639771 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508194 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14763336 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14522866 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508202 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508155 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508174 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508212 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32508163 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10600705 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600706 - -
          Affymetrix Mouse Gene 1.0 ST Array 10600704 - -
          Affymetrix Mouse Gene 1.1 ST Array 10600703 - -
          Mouse Genome 430 2.0 Array 1449985_PM_at - -
          Mouse Genome 430 2.0 Array 1449985_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73495_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73495_at - -
          Affymetrix Mouse Clariom S Array PSR0X00005560.mm.1 NM_028554.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005560.mm.1 NM_028554.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005560.mm.1 TC0X00000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005559.mm.1 NM_028554.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005559.mm.1 NM_028554.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005559.mm.1 TC0X00000843.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00005558.mm.1 NM_028554.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005558.mm.1 NM_028554.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00005558.mm.1 TC0X00000843.mm.1 MTA TC
          Mouse Expression Set 430 1449985_at - -
          Mouse Expression Set 430 1449985_PM_at - -
          Murine Genome U74v2 Set 168903_PM_at - -
          Murine Genome U74v2 Set 168903_at - -
          Mouse Genome 430A 2.0 Array 1449985_at - -
          Mouse Genome 430A 2.0 Array 1449985_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 560815 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1410551 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001410551 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2593059 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2593059 - -
          mouse-6 v1.1 (Illumina) scl54795.3.1_10-S - -
          Illumina Sentrix mouse-6.v1 bead array 1410551 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl54795.3.1_10-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1410551 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160066 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3080550 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2593058 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2593058 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3080550 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160066 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-mrft7:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0