Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03031408_s1
          See other PIH1D3 GE Assays ›
          Gene Symbol:
          Pih1d3
          Gene Name:
          PIH1 domain containing 3
          Gene Aliases:
          4930521A18Rik
          Chromosome Location:
          Chr.1: 31222839 - 31224287 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03031408_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pih1d3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_029062.2 NP_083338.1 1 851 82
          GenBank mRNA AK015858.1 - 1 853 82
          BC107262.2 - 1 843 40052516 82

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pih1d3

          Gene Ontology Categories:

          Function(s) Process(es)

          sperm motility
          axonemal dynein complex assembly
          protein binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534001 AK015858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534001 NM_029062.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534001 AK015858.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P534001 NM_029062.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 AB046173.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 AK011689.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 AK011797.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 AK030096.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 AK088465.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 AK147140.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 BC045138.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 BC049082.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 626722 NM_023670.2 refseq_mrna_mouse_NM
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534001 AK015858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534001 NM_029062.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534001 AK015858.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P534001 NM_029062.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123435 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123445 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123450 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123429 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123438 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123422 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123443 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123451 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123449 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123426 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123454 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123428 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123453 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123447 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123423 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123442 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123437 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123420 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123433 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123431 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123436 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123441 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123418 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P20123424 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00037099 - -
          Whole Mouse Genome G4122A A_52_P534001 AK015858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534001 NM_029062.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534001 AK015858.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P534001 NM_029062.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00701400 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00701416 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00701412 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00701411 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P00701414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123461 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123416 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123466 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123426 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123486 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123488 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123478 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123435 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123428 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123445 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123484 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P20123457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123454 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123438 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123435 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123441 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00701402 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123445 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00321007 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123423 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00220495 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123461 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123449 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123442 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123451 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123431 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123450 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00548350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123456 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123418 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123447 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123426 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P00366898 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123433 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123463 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123466 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123487 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P20123459 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10345198 - -
          Affymetrix Mouse Gene 1.0 ST Array 10345199 - -
          Affymetrix HT MG-430 PM Array Plate 1430400_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10345197 - -
          Mouse Genome 430 2.0 Array 1430400_PM_at - -
          Mouse Genome 430 2.0 Array 1430400_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74708_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 74708_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0100001180.mm.1 NM_029062.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001180.mm.1 NM_029062.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001180.mm.1 TC0100000194.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0100001181.mm.1 NM_029062.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001181.mm.1 NM_029062.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0100001181.mm.1 TC0100000194.mm.1 MTA TC
          Mouse Expression Set 430 1430400_at - -
          Mouse Expression Set 430 1430400_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430400_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430400_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 382448 - -
          ABI Mouse Genome Survey Microarray 854691 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2524046 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2645001 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2645001 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2980560 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2980560 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2qnx8:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0