Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03032137_s1
          See other COA4 GE Assays ›
          Gene Symbol:
          Coa4
          Gene Name:
          cytochrome c oxidase assembly factor 4
          Gene Aliases:
          5330414O08Rik, AI115477, AW047259, Chchd8, E2ig2
          Chromosome Location:
          Chr.7: 100537100 - 100539812 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03032137_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Coa4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_183270.2 NP_899093.1 2 269 104
          GenBank mRNA AK008883.1 - 2 235 104
          AK019895.1 - 241 104
          AK088786.1 - 2 257 104
          AK161506.1 - 270 104
          BC048574.1 - 2 210 104
          BC116995.1 - 2 238 40126314 104
          BC116997.1 - 2 238 40126316 104

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Coa4

          Panther Classification:

          Molecular Function -

          chaperone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368309 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368311 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368312 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368310 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091631 BC048574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091631 BC096605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091631 AK161506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091631 NM_183270.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2091631 AK008883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 BC048574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 BC096605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 AK161506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 NM_183270.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 AK008883.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 BC048574.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 BC096605.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 AK161506.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 NM_183270.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P434841 AK008883.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 625141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687671 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 968876 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 44828 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P101827 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091631 BC048574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091631 BC096605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091631 AK161506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091631 NM_183270.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2091631 AK008883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 BC048574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 BC096605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 AK161506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 NM_183270.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 AK008883.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 BC048574.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 BC096605.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 AK161506.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 NM_183270.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P434841 AK008883.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368328 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368321 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368320 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368318 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368300 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368316 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368304 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368322 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368312 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368289 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368323 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368327 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368325 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368287 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368309 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368324 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368306 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368313 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368301 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368308 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25368285 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P101827 - -
          Whole Mouse Genome G4122A A_52_P434841 BC048574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 BC096605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 AK161506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 NM_183270.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 AK008883.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 BC048574.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 BC096605.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 AK161506.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 NM_183270.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P434841 AK008883.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P125386 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P101827 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06025907 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06025906 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06025904 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06122661 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368310 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368311 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368309 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25368312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368300 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368328 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368349 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368332 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368264 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368299 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368324 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368304 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25368275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368322 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368329 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368324 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368318 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06601510 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368292 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368264 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06025900 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05974834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368291 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368336 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06437581 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06378654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368343 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368303 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06025906 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368335 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06601516 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368321 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368301 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06284134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368289 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368313 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368328 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368332 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368338 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368316 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05955106 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06470569 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06378645 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368348 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368306 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368275 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06039085 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06248298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P25368302 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10555410 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555409 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555408 - -
          Affymetrix Mouse Gene 1.0 ST Array 10555411 - -
          Affymetrix HT MG-430 PM Array Plate 1430540_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10555407 - -
          Mouse Genome 430 2.0 Array 1430540_at - -
          Mouse Genome 430 2.0 Array 1430540_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700012056.mm.1 TC0700001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012056.mm.1 NM_183270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012056.mm.1 NM_183270.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012048.mm.1 TC0700001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012048.mm.1 NM_183270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012048.mm.1 NM_183270.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012055.mm.1 TC0700001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012055.mm.1 NM_183270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012055.mm.1 NM_183270.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012050.mm.1 TC0700001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012050.mm.1 NM_183270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012050.mm.1 NM_183270.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012054.mm.1 TC0700001410.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700012054.mm.1 NM_183270.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700012054.mm.1 NM_183270.2 Refseq
          Mouse Expression Set 430 1430540_at - -
          Mouse Expression Set 430 1430540_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1430540_at - -
          Affymetrix HT Mouse Genome 430B Array 1430540_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 507807 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3800088 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003800088 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1218282 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1218282 - -
          mouse-6 v1.1 (Illumina) scl32313.5_285-S - -
          Illumina Sentrix mouse-6.v1 bead array 3800088 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl32313.5_285-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3800088 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2817151 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1218282 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2817151 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline