Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03033061_u1
          See other LOC547349 GE Assays ›
          Gene Symbol:
          LOC547349
          Gene Name:
          MHC class I family member
          Gene Aliases:
          -
          Chromosome Location:
          -
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03033061_u1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: LOC547349
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001025208.1 NP_001020379.1 51 99
          GenBank mRNA AY989869.1 - 51 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Recommended Assay Mus musculus

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene LOC547349

          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966659 NM_001025208.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966659 AY533669.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966659 AY989869.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966660 NM_001025208.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966660 AY533669.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1966660 AY989869.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 813957 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 316141 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615809 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 784380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 563044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572196 AK048053.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572196 BC039781.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572196 BC066195.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572196 BC098506.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572196 BC116304.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572196 BC116305.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 572196 NM_001029982.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 809110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 264921 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425171 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 401180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 799766 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 899818 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 459471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533592 AK042148.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912597 AK036531.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912597 AK054132.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912597 AK086904.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912597 AK136577.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912597 AK137515.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912597 BC005440.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 912597 NM_008964.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 732128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 852509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558327 AF017631.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558327 AK164358.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558327 NM_008989.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558327 U02098.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558327 XM_001475407.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558327 XM_001475421.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 558327 XM_001475437.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 835151 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 238570 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 269547 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 26919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 103181 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 135530 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 66593 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157931 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966659 NM_001025208.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966659 AY533669.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966659 AY989869.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966660 NM_001025208.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966660 AY533669.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1966660 AY989869.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08768025 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136941 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08768095 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08768112 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03311704 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03311716 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08768074 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08768107 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03311702 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P158147 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136941 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P158147 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08768095 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08768025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158605 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158812 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158809 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158813 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158780 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31158610 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158637 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158670 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158909 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158674 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158663 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13055602 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158667 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158653 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158660 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13542011 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13421927 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13539342 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13542010 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158910 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158639 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13122457 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31158658 - -
          Array Name Probe IDs Targets Target Type
          Mouse Genome 430 2.0 Array 1447260_PM_at - -
          Mouse Genome 430 2.0 Array 1447260_at - -
          Mouse Expression Set 430 1447260_PM_at - -
          Mouse Expression Set 430 1447260_at - -
          Murine Genome U74v2 Set 102161_f_at - -
          Murine Genome U74v2 Set 102161_PM_f_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2732816 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2732816 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline