| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400015206.mm.1 |
NM_001081248.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400015206.mm.1 |
NM_001081248.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400015206.mm.1 |
TC0400001854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400015205.mm.1 |
TC0400001854.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400015207.mm.1 |
NM_001081248.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400015207.mm.1 |
NM_001081248.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400015207.mm.1 |
TC0400001854.mm.1 |
MTA TC |