Search
Search

Dgcr14| Gene Symbol: Dgcr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081633.1 | NP_001075102.1 | 5 - 6 | 743 | 88 | |
| NM_022408.2 | NP_071853.2 | 5 - 6 | 746 | 88 | ||
| GenBank mRNA | AF037256.1 | - | 5 - 6 | 745 | 88 | |
| AK088520.1 | - | 5 - 6 | 708 | 88 | ||
| AK142684.1 | - | 5 - 6 | 711 | 88 | ||
| AK148404.1 | - | 5 - 6 | 714 | 88 | ||
| BC013711.1 | - | 5 - 6 | 722 | 3708646 | 88 | |
Set Membership: |
|