Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03038703_s1
          See other FAM43B GE Assays ›
          Gene Symbol:
          Fam43b
          Gene Name:
          family with sequence similarity 43, member B
          Gene Aliases:
          OTTMUSG00000009974
          Chromosome Location:
          Chr.4: 138394092 - 138396458 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03038703_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam43b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001081672.2 NP_001075141.1 1 786 153
          GenBank mRNA AK134570.1 - 1 786 153

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998591 BB677246.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1998591 NM_001081672.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 845079 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 79356 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998591 BB677246.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1998591 NM_001081672.2 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345562 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345553 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345500 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345535 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345508 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345545 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345506 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345549 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345566 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345592 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345537 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345509 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345503 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345570 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345526 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345584 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345558 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345568 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345499 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345582 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345528 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345579 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345559 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345565 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23345590 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03478884 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345575 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03995981 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345573 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345578 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03459631 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03214381 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345569 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03913792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03478889 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03913793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345548 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03261618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345567 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345547 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03266303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345529 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345583 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345581 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345542 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345549 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345566 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03478887 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285440 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345531 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345559 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03259844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03913788 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345536 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345570 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285439 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345550 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345584 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345587 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345519 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345528 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03266304 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345579 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345593 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03285436 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23345551 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10517615 - -
          Affymetrix HT MG-430 PM Array Plate 1435063_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10517614 - -
          Mouse Genome 430 2.0 Array 1435063_PM_at - -
          Mouse Genome 430 2.0 Array 1435063_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 625638_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 625638_at - -
          Affymetrix Mouse Clariom S Array PSR0400031507.mm.1 TC0400003823.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400031507.mm.1 NM_001081672.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400031507.mm.1 NM_001081672.2 Refseq
          Mouse Expression Set 430 1435063_at - -
          Mouse Expression Set 430 1435063_PM_at - -
          Murine Genome U74v2 Set 140621_PM_at - -
          Murine Genome U74v2 Set 140621_at - -
          Affymetrix HT Mouse Genome 430B Array 1435063_at - -
          Affymetrix HT Mouse Genome 430B Array 1435063_PM_at - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline