| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1983044 |
NM_001081687.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1983044 |
DQ055451.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
130074 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
922971 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
729826 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
152965 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
857311 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
122551 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
1265 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK011233.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK012824.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK019371.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK029431.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK032206.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK075716.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK147795.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
AK148333.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
BC012271.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
BC019795.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
BC030190.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
911379 |
NM_026390.2 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
534900 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
841932 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
522999 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829581 |
AK131853.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829581 |
BC039973.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829581 |
XM_001472076.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829581 |
XM_001475252.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829581 |
XM_001476886.1 |
refseq_mrna_mouse_XM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
917345 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
579415 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
402644 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
539875 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
229783 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
243396 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
924618 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
91197 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
6425 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
66553 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
30632 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
106323 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
27486 |
- |
- |
| Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P140969 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1983044 |
NM_001081687.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1983044 |
DQ055451.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P465129 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961750 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961925 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961887 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961774 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961930 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961896 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961809 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961819 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961872 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961815 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961711 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961735 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961718 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961710 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961771 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961879 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961705 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961757 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961873 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961898 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961858 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961920 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961749 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961811 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961860 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961875 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961888 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961725 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961911 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961931 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961917 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961743 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961724 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961838 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961822 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961892 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961722 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961923 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961715 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961842 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961919 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961882 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961813 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961894 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961821 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961929 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961707 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961726 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961900 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961899 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961928 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961776 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961753 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961840 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961891 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961817 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961933 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961807 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961874 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961885 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961728 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961803 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P21961818 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961911 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961916 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961931 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961928 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961912 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961920 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961917 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961929 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961933 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961923 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961919 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02460357 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961925 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02870645 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P21961926 |
- |
- |