Search
Search

Wfdc8| Gene Symbol: Wfdc8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080550.2 | NP_001074019.1 | 3 - 4 | 632 | 68 | |
| NM_001276431.1 | NP_001263360.1 | 3 - 4 | 632 | 68 | ||
| NM_001276432.1 | NP_001263361.1 | 3 - 4 | 1461 | 68 | ||
| GenBank mRNA | AK136599.1 | - | 3 - 4 | 1462 | 68 | |
| AY669506.1 | - | 3 - 4 | 632 | 68 | ||
| AY757956.1 | - | 3 - 4 | 632 | 68 | ||
Set Membership: |
|