Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03039419_m1
          See other B230217C12RIK GE Assays ›
          Gene Symbol:
          B230217C12Rik
          Gene Name:
          RIKEN cDNA B230217C12 gene
          Gene Aliases:
          AI840637
          Chromosome Location:
          Chr.11: 97840780 - 97843043 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03039419_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: B230217C12Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080935.1 NP_001074404.1 1 - 2 332 79
          GenBank mRNA AK082773.1 - 1 - 2 262 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001551.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK082773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK042827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK080808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK081899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK018341.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK082773.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK042827.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK080808.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK081899.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P403246 AK018341.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 566364 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P146314 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK082773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK042827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK080808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK081899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK018341.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK082773.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK042827.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK080808.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK081899.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P403246 AK018341.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P146314 - -
          Whole Mouse Genome G4122A A_52_P208787 AK082773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208787 AK140210.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208787 AK042827.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208787 AK080808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P208787 AK081899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK082773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK042827.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK080808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK081899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK018341.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK082773.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK042827.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK080808.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK081899.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P403246 AK018341.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105489 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P146314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053064 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053036 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053015 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28053017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053060 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053017 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09379342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053050 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053047 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09523859 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09249214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053015 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09339713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053063 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09646068 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09379331 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053012 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053048 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28053036 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10380831 - -
          Affymetrix Mouse Gene 1.0 ST Array 10380832 - -
          Affymetrix HT MG-430 PM Array Plate 1428568_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1428569_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10380830 - -
          Mouse Genome 430 2.0 Array 1428569_PM_at - -
          Mouse Genome 430 2.0 Array 1428568_PM_at - -
          Mouse Genome 430 2.0 Array 1428569_at - -
          Mouse Genome 430 2.0 Array 1428568_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68127_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68127_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100014324.mm.1 NM_001080935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014324.mm.1 NM_001080935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014324.mm.1 TC1100001551.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100014327.mm.1 NM_001080935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014327.mm.1 NM_001080935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014327.mm.1 TC1100001551.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100014320.mm.1 NM_001080935.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014320.mm.1 NM_001080935.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100014320.mm.1 TC1100001551.mm.1 MTA TC
          Mouse Expression Set 430 1428568_PM_at - -
          Mouse Expression Set 430 1428569_PM_at - -
          Mouse Expression Set 430 1428569_at - -
          Mouse Expression Set 430 1428568_at - -
          Affymetrix HT Mouse Genome 430B Array 1428569_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428568_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428568_at - -
          Affymetrix HT Mouse Genome 430B Array 1428569_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 913925 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 101940435 - -
          MouseWG-6 v2 Expression BeadChip 105860347 - -
          MouseWG-6 v2 Expression BeadChip 101240195 - -
          Illumina mouse-6 v1.0 expression beadchip 104590717 - -
          Illumina mouse-6 v1.0 expression beadchip 105860347 - -
          Illumina mouse-6 v1.0 expression beadchip 101940435 - -
          Illumina mouse-6 v1.0 expression beadchip 102320064 - -
          Illumina mouse-6 v1.0 expression beadchip 104610373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227674 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1215777 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237335 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1252991 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1256158 - -
          Illumina Sentrix mouse-6.v1 bead array 105860347 - -
          Illumina Sentrix mouse-6.v1 bead array 101940435 - -
          Illumina Sentrix mouse-6.v1 bead array 104610373 - -
          Illumina Sentrix mouse-6.v1 bead array 104590717 - -
          Illumina Sentrix mouse-6.v1 bead array 102320064 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 105860347 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 101940435 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104590717 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104610373 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 102320064 - -
          MouseRef-8 v2 Expression BeadChip ILMN_3040617 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1256158 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1252991 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237335 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3040617 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline