Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03039759_s1
          See other RNF208 GE Assays ›
          Gene Symbol:
          Rnf208
          Gene Name:
          ring finger protein 208
          Gene Aliases:
          1110061N23Rik
          Chromosome Location:
          Chr.2: 25242929 - 25244261 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03039759_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf208
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_176834.2 NP_789804.2 1 936 87
          GenBank mRNA AK004344.1 - 1 160 87
          BC030073.1 - 2 738 5254639 87
          BQ031423.1 - 1 87

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf208

          Gene Ontology Categories:

          Function(s) Process(es)

          protein autoubiquitination
          ubiquitin-protein transferase activity
          zinc ion binding
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052281 NM_176834.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052281 BC030073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2052281 BQ031423.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035812 NM_176834.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2035812 BQ031423.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 757856 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137220 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035812 NM_176834.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2035812 BQ031423.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052281 NM_176834.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052281 BC030073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2052281 BQ031423.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070820 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070810 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070817 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070814 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070813 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070824 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070811 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070822 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21070819 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P137220 - -
          Whole Mouse Genome G4122A A_51_P217123 NM_176834.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P217123 AK004344.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P217123 BC030073.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P127195 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P137220 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01878259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070811 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070892 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070819 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070900 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070822 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070735 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070890 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070789 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070871 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070875 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070801 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070739 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21070726 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02023972 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02023975 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02023970 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02023971 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070801 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02023964 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21070805 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10469952 - -
          Affymetrix Mouse Gene 1.0 ST Array 10469953 - -
          Affymetrix HT MG-430 PM Array Plate 1435105_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10469951 - -
          Mouse Genome 430 2.0 Array 1435105_PM_at - -
          Mouse Genome 430 2.0 Array 1435105_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68846_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 68846_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200002222.mm.1 NM_176834.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002222.mm.1 NM_176834.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200002222.mm.1 TC0200000403.mm.1 MTA TC
          Mouse Expression Set 430 1435105_at - -
          Mouse Expression Set 430 1435105_PM_at - -
          Murine Genome U74v2 Set 103771_PM_at - -
          Murine Genome U74v2 Set 103771_at - -
          Mouse Genome 430A 2.0 Array 1435105_at - -
          Mouse Genome 430A 2.0 Array 1435105_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435105_at - -
          Affymetrix HT Mouse Genome 430A Array 1435105_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 488486 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 3360692 - -
          Illumina mouse-6 v1.0 expression beadchip 3360692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003360692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2636349 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2636349 - -
          mouse-6 v1.1 (Illumina) scl21188.2_9-S - -
          Illumina Sentrix mouse-6.v1 bead array 3360692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl21188.2_9-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3360692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2636349 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2636349 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl21188.2_9-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline