Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03040519_sH
          See other OLFR322 GE Assays ›
          Gene Symbol:
          Olfr322
          Gene Name:
          olfactory receptor 322
          Gene Aliases:
          GA_x5J8B7U1E65-1-838, MOR256-64, OTTMUSG00000005755, Olfr179, Olfr179-ps1, Olfr216, Olfr216-ps1, Olfr322-ps1
          Chromosome Location:
          Chr.11: 58665561 - 58666544 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03040519_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr322
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_207693.1 NP_997576.1 1 631 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P353072 NM_207693.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P353072 NM_207693.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832240 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27832198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09150193 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09685563 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832172 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832162 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832249 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09150173 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832217 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09030652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832197 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832188 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09150199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09591149 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832195 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09150190 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832177 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09685360 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27832199 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10376403 - -
          Affymetrix Mouse Gene 1.1 ST Array 10376402 - -
          Affymetrix Mouse Clariom S Array PSR1100005588.mm.1 NM_207693.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005588.mm.1 NM_207693.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100005588.mm.1 TC1100000696.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2947039 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2947039 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline