Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03040564_gH
          See other GM5382 GE Assays ›
          Gene Symbol:
          Gm5382
          Gene Name:
          predicted gene 5382
          Gene Aliases:
          EG385328
          Chromosome Location:
          Chr.X: 14211148 - 14211657 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03040564_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm5382
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001034100.1 NP_001029272.1 1 449 112
          GenBank mRNA BC100482.1 - 1 449 30918325 112

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          histone

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P221588 NM_001034100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P221588 BC100482.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P221588 NM_001034100.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P221588 BC100482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P221588 NM_001034100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P221588 BC100482.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P221588 NM_001034100.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P221588 BC100482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P221588 NM_001034100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P221588 BC100482.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P221588 NM_001034100.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P221588 BC100482.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15182664 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P14840043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269519 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269518 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269460 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269546 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269477 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269544 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269459 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269456 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269539 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269464 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269524 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269523 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269547 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269514 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269538 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269462 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269455 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269448 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269536 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269482 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269450 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269469 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269521 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269529 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269527 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269512 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269487 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269490 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269479 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269468 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269436 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269443 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269476 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269511 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269474 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269485 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269449 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269533 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269471 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269454 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269452 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269525 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269481 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269458 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269465 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269543 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32269478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269441 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269517 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269462 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14910123 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269482 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269477 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269456 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15182455 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269452 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14522601 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269518 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269458 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269459 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14840043 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P15182470 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269536 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269539 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269479 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14869303 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14526407 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269524 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269525 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14632816 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269484 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269474 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269511 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269478 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14910104 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269519 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14875841 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269543 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269454 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269448 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269481 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269533 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269521 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269526 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269507 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269465 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269469 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269443 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269529 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269523 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269485 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P14910121 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269489 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269464 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269468 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269453 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269544 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269541 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269514 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269548 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269546 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269471 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269490 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269540 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269509 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32269444 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10598764 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598593 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598647 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598597 - -
          Affymetrix Mouse Gene 1.0 ST Array 10598653 - -
          Affymetrix Mouse Gene 1.1 ST Array 10598763 - -
          Affymetrix Mouse Clariom S Array PSR0X00001115.mm.1 NM_001034100.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001115.mm.1 NM_001034100.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00001115.mm.1 TC0X00000172.mm.1 MTA TC
          Murine Genome U74v2 Set 169679_PM_at - -
          Murine Genome U74v2 Set 169679_at - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2854888 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2854888 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline