Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03047263_m1
          See other NLRP1A GE Assays ›
          Gene Symbol:
          Nlrp1a
          Gene Name:
          NLR family, pyrin domain containing 1A
          Gene Aliases:
          CARD7, DEFCAP, Gm14, Gm15, NAC, Nalp1, Nalp1a, Nlrp1, PP1044
          Chromosome Location:
          Chr.11: 71091197 - 71144704 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03047263_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Nlrp1a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001004142.2 NP_001004142.2 2 - 3 365 73
          GenBank mRNA AK157034.1 - 2 - 3 366 73
          AY355339.1 - 1 - 2 147 73
          DQ117601.1 - 1 - 2 306 73
          KC539856.1 - 1 - 2 147 73
          KC539857.1 - 1 - 2 147 73
          KC539858.1 - 1 - 2 147 73
          KC539859.1 - 1 - 2 147 73
          KC539860.1 - 1 - 2 147 73
          KC539861.1 - 1 - 2 147 73

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003124.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Nlrp1a

          Gene Ontology Categories:

          Function(s) Process(es)

          response to muramyl dipeptide
          defense response to bacterium
          positive regulation of interleukin-1 beta secretion
          programmed necrotic cell death
          protein binding
          scaffold protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023146 DQ117601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023146 AY355339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023146 NM_001004142.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2070366 AY355339.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955998 AK041714.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955998 BC064077.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955998 BC038128.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955998 BC059731.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1955998 AK002779.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 146259 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727286 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509128 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 200038 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 136139 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 232934 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 930395 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850698 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 415514 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 833795 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 105772 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 533796 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 12456 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 808538 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829592 AK142371.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829592 BC056625.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829592 BC081546.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 829592 NM_009660.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 677199 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 247860 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 102180 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 743961 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509602 AK078998.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509602 AK085392.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509602 BC061041.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509602 NM_001045554.1 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509602 NM_138685.2 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540360 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 290890 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 966744 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 687326 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 125734 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 161413 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 78489 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 150013 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27280 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 14966 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023146 DQ117601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023146 AY355339.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023146 NM_001004142.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955998 AK041714.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955998 BC064077.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955998 BC038128.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955998 BC059731.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1955998 AK002779.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P253757 DQ117601.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P253757 AY355339.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P253757 NM_001004142.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904372 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904378 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904386 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904369 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904381 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904396 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904388 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904400 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904373 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904375 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904391 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904393 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904399 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904392 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904370 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904383 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27904382 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07390322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07390389 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07390410 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02358482 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07390331 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02358460 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07390374 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02358483 - -
          Whole Mouse Genome G4122A A_52_P253757 DQ117601.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P253757 AY355339.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P253757 NM_001004142.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07390331 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07390410 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P07390389 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09372622 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09207411 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09372617 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09372625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904286 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904384 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904306 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904200 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904289 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904315 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904249 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904361 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904309 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904355 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27904364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904394 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09267549 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09270639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904380 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09171618 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904375 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09372625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904366 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904369 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09649187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904391 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904386 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09228878 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904373 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904378 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904364 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09249123 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904396 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904388 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904402 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904392 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904384 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27904361 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10388063 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388051 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388055 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388052 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388053 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388049 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388054 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388048 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388061 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388050 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388059 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388060 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388058 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388064 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388057 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388062 - -
          Affymetrix Mouse Gene 1.0 ST Array 10388056 - -
          Affymetrix Mouse Gene 1.1 ST Array 10388047 - -
          Affymetrix Mouse Clariom S Array PSR1100028955.mm.1 NM_001004142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028955.mm.1 NM_001004142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028955.mm.1 TC1100003124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028958.mm.1 NM_001004142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028958.mm.1 NM_001004142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028958.mm.1 TC1100003124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028956.mm.1 NM_001004142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028956.mm.1 NM_001004142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028956.mm.1 TC1100003124.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100028957.mm.1 NM_001004142.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028957.mm.1 NM_001004142.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100028957.mm.1 TC1100003124.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 555754 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160230 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160230 - -
          mouse-6 v1.1 (Illumina) GI_51921280-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160230 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160230 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline