Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03047635_s1
          See other OLFR920 GE Assays ›
          Gene Symbol:
          Olfr920
          Gene Name:
          olfactory receptor 920
          Gene Aliases:
          MOR165-6
          Chromosome Location:
          Chr.9: 38752796 - 38756863 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03047635_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr920
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146787.2 NP_666998.2 2 1527 151
          GenBank mRNA AK040176.1 - 1 674 151

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr920

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085200 AK040176.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2085200 NM_146787.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181936 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 527054 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P138074 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085200 AK040176.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2085200 NM_146787.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P196581 AK040176.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P196581 NM_146787.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435967 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435950 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435960 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435944 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435968 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435957 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435939 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435949 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435972 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435958 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435946 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435952 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435959 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435966 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435943 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435973 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435948 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435955 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435962 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435964 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435953 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435942 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435940 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435956 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435965 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26435971 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P138074 - -
          Whole Mouse Genome G4122A A_51_P196581 AK040176.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P196581 NM_146787.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123860 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P138074 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07926040 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07926037 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07926034 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08052338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07926038 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07926028 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435952 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435949 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436001 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435966 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436004 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435972 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436021 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435956 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435986 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435932 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435955 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435950 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436003 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435965 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26436010 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435934 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435944 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435922 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435973 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435962 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435964 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26435939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435952 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435948 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08110678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435986 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435964 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435996 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435976 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435965 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435950 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435972 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435994 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435995 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435940 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435981 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435942 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435934 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435956 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435999 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435953 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435932 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435962 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435949 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435955 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435926 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435966 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435935 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435938 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435933 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435925 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435946 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435944 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436001 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26436003 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26435984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07926033 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10584459 - -
          Affymetrix Mouse Gene 1.1 ST Array 10584458 - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 783565 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2470242 - -
          Illumina mouse-6 v1.0 expression beadchip 2470242 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002470242 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232797 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232797 - -
          mouse-6 v1.1 (Illumina) scl37085.1.1_60-S - -
          Illumina Sentrix mouse-6.v1 bead array 2470242 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl37085.1.1_60-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2470242 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1232797 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232797 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline