Search
Search

Rabgef1| Gene Symbol: Rabgef1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199059.1 | NP_001185988.1 | 3 - 4 | 427 | 70 | |
| NM_019983.2 | NP_064367.1 | 2 - 3 | 572 | 70 | ||
| GenBank mRNA | AF093590.1 | - | 3 - 4 | 403 | 70 | |
| AK044568.1 | - | 2 - 3 | 592 | 70 | ||
| AK146734.1 | - | 3 - 4 | 377 | 70 | ||
| BC018229.1 | - | 2 - 3 | 572 | 4507477 | 70 | |
Set Membership: |
|