Search
Search

Krtap12-1| Gene Symbol: Krtap12-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010670.1 | NP_034800.1 | 1 | 255 | 82 | |
| GenBank mRNA | BC107319.1 | - | 1 | 242 | 40053395 | 82 |
| BC107320.1 | - | 1 | 242 | 40053396 | 82 | |
Set Membership: |
|