Search
Search

Olfr1446| Gene Symbol: Olfr1446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146704.1 | NP_666915.1 | 1 | 442 | 76 | |
| GenBank mRNA | BC119061.1 | - | 1 | 525 | 8733810 | 76 |
| BC137821.1 | - | 1 | 525 | 8860840 | 76 | |
Set Membership: |
|