Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03052303_s1
          See other PSG28 GE Assays ›
          Gene Symbol:
          Psg28
          Gene Name:
          pregnancy-specific glycoprotein 28
          Gene Aliases:
          cea16
          Chromosome Location:
          Chr.7: 18422536 - 18432055 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03052303_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psg28
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_054063.4 NP_473404.3 5 1651 139
          GenBank mRNA AF113598.1 - 5 1501 139
          BC099402.1 - 5 1637 30787049 139
          BU754932.1 - 1 139

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002452.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psg28

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24988531 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100186 BC099403.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100186 BC141038.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100186 CK020635.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100186 NM_054063.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100186 AF113598.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074521 BU754932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074521 NM_054063.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074521 AF113598.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074521 BC099402.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 241567 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 948370 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84251 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 308912 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 255327 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 607865 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246597 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283380 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368710 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 615465 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 658392 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672848 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390811 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 367435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 249509 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 13548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8362 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 137270 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 108325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 156064 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52224 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 7034 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074521 BU754932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074521 NM_054063.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074521 AF113598.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074521 BC099402.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100186 BC099403.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100186 BC141038.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100186 CK020635.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100186 NM_054063.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100186 AF113598.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24988619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24988666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24988635 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06068645 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24988531 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24988580 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24988541 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24988551 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24988548 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24988666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24988570 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05987243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24988619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24988635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24988666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06145026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06277034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06145031 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10560369 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560368 - -
          Affymetrix Mouse Gene 1.0 ST Array 10560367 - -
          Affymetrix HT MG-430 PM Array Plate 1425881_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10560366 - -
          Mouse Genome 430 2.0 Array 1425881_at - -
          Mouse Genome 430 2.0 Array 1425881_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114871_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 114871_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700021586.mm.1 TC0700002452.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021586.mm.1 NM_054063.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021586.mm.1 NM_054063.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021587.mm.1 TC0700002452.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700021587.mm.1 NM_054063.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700021587.mm.1 NM_054063.4 Refseq
          Mouse Expression Set 430 1425881_PM_at - -
          Mouse Expression Set 430 1425881_at - -
          Murine Genome U74v2 Set 167203_at - -
          Murine Genome U74v2 Set 167203_PM_at - -
          Mouse Genome 430A 2.0 Array 1425881_PM_at - -
          Mouse Genome 430A 2.0 Array 1425881_at - -
          Affymetrix HT Mouse Genome 430A Array 1425881_at - -
          Affymetrix HT Mouse Genome 430A Array 1425881_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 484584 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4070128 - -
          Illumina mouse-6 v1.0 expression beadchip 4070128 - -
          Illumina mouse-6 v1.0 expression beadchip 104760333 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004070128 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2627050 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2627050 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2542630 - -
          mouse-6 v1.1 (Illumina) scl00114871.1_73-S - -
          mouse-6 v1.1 (Illumina) GI_31340566-S - -
          Illumina Sentrix mouse-6.v1 bead array 104760333 - -
          Illumina Sentrix mouse-6.v1 bead array 4070128 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00114871.1_73-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 104760333 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4070128 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2627050 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2627050 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl00114871.1_73-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline