Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03052681_sH
          See other UGT2B38 GE Assays ›
          Gene Symbol:
          Ugt2b38
          Gene Name:
          UDP glucuronosyltransferase 2 family, polypeptide B38
          Gene Aliases:
          9430041C03Rik, AA986709
          Chromosome Location:
          Chr.5: 87409939 - 87424203 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03052681_sH
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ugt2b38
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133894.2 NP_598655.1 6 1662 147
          GenBank mRNA AK034801.1 - 6 1663 147
          BC015272.1 - 6 1659 4241480 147
          BC057169.1 - 6 1648 4975249 147
          BC069923.1 - 6 1650 4974994 147

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ugt2b38

          Gene Ontology Categories:

          Function(s) Process(es)

          flavonoid biosynthetic process
          cellular response to hormone stimulus
          flavonoid glucuronidation
          glucuronosyltransferase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131389 BC069923.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131389 AK034801.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131389 BC015272.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131389 BC057169.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P131389 NM_133894.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 49134 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 862463 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 89230 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239646 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 149157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 365205 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 965782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 93925 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 40232 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131389 BC069923.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131389 AK034801.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131389 BC015272.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131389 BC057169.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P131389 NM_133894.2 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867767 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867764 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867745 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867758 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867761 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867763 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867746 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867759 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867765 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23867768 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05644762 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05644724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01153842 - -
          Whole Mouse Genome G4122A A_52_P35340 BC069923.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P35340 AK034801.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P35340 BC015272.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P35340 BC057169.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P35340 NM_133894.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P01153842 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867730 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867728 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23867674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867722 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867730 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04981568 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867638 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867768 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867763 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04510122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04444618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867729 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867650 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867764 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867765 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867759 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867746 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04258787 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867735 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04744953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867737 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867645 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867761 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04744954 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04360372 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867711 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23867724 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10531076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10531077 - -
          Affymetrix HT MG-430 PM Array Plate 1423397_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10531073 - -
          Mouse Genome 430 2.0 Array 1423397_at - -
          Mouse Genome 430 2.0 Array 1423397_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100559_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 100559_PM_at - -
          Mouse Expression Set 430 1423397_PM_at - -
          Mouse Expression Set 430 1423397_at - -
          Murine Genome U74v2 Set 137212_PM_f_at - -
          Murine Genome U74v2 Set 137211_PM_i_at - -
          Murine Genome U74v2 Set 137211_i_at - -
          Murine Genome U74v2 Set 137212_f_at - -
          Mouse Genome 430A 2.0 Array 1423397_PM_at - -
          Mouse Genome 430A 2.0 Array 1423397_at - -
          Affymetrix HT Mouse Genome 430A Array 1423397_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1423397_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 669550 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1570341 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001570341 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2730424 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2730424 - -
          mouse-6 v1.1 (Illumina) scl0100559.1_228-S - -
          Illumina Sentrix mouse-6.v1 bead array 1570341 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0100559.1_228-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1570341 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2730424 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2730424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline