Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03053368_s1
          See other TEX19.1 GE Assays ›
          Gene Symbol:
          Tex19.1
          Gene Name:
          testis expressed gene 19.1
          Gene Aliases:
          2410081M02Rik, Tex19, Tex19a, mTex19.1
          Chromosome Location:
          Chr.11: 121146143 - 121148314 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03053368_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tex19.1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028602.2 NP_082878.2 3 1657 153
          GenBank mRNA AF285590.1 - 3 1657 153
          AK010735.1 - 3 1640 153
          BC053492.1 - 3 1626 30024269 159

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tex19.1

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          placenta development
          synapsis
          spermatogenesis
          male gonad development
          negative regulation of transposition
          posttranscriptional regulation of gene expression
          sexual reproduction
          cell differentiation
          meiotic cell cycle
          spongiotrophoblast cell proliferation
          cell proliferation involved in embryonic placenta development
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245533 AK010735.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P245533 AK010735.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928593 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179217 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245533 AK010735.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P245533 AK010735.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188866 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188874 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188845 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188853 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188882 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188847 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188843 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188880 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188890 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188846 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188849 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188870 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188858 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188850 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188884 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188885 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188887 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188889 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188878 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188863 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28188860 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179217 - -
          Whole Mouse Genome G4122A A_51_P245533 AK010735.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P245533 AK010735.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105594 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179217 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08974892 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09484772 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09484771 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08974887 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P08974889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188919 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188874 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188928 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188880 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188945 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188896 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188847 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188921 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188936 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188925 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188938 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188836 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188913 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188834 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188864 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188930 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188840 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188833 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188878 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188824 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188839 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188895 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188940 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188853 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188849 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188887 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28188892 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188885 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188853 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188825 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188839 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188847 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188887 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09242676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188860 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188843 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09071461 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188882 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188834 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188836 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188874 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09242678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09242679 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188840 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09548660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08974889 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09242680 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09676989 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09093596 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340436 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08974896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188896 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09484771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188856 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188898 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09340440 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188895 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09629075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188866 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188846 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28188827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09093595 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10383512 - -
          Affymetrix Mouse Gene 1.0 ST Array 10383513 - -
          Affymetrix HT MG-430 PM Array Plate 1417482_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10383511 - -
          Mouse Genome 430 2.0 Array 1417482_PM_at - -
          Mouse Genome 430 2.0 Array 1417482_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73679_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73679_at - -
          Mouse Expression Set 430 1417482_PM_at - -
          Mouse Expression Set 430 1417482_at - -
          Murine Genome U74v2 Set 102418_PM_at - -
          Murine Genome U74v2 Set 102418_at - -
          Mouse Genome 430A 2.0 Array 1417482_PM_at - -
          Mouse Genome 430A 2.0 Array 1417482_at - -
          Affymetrix HT Mouse Genome 430A Array 1417482_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417482_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 598084 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 6040114 - -
          Illumina mouse-6 v1.0 expression beadchip 6040114 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006040114 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2615917 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2615917 - -
          mouse-6 v1.1 (Illumina) scl40619.3_467-S - -
          Illumina Sentrix mouse-6.v1 bead array 6040114 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40619.3_467-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6040114 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2615917 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2615917 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline