Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03057690_m1
          See other GM436 GE Assays ›
          Gene Symbol:
          Gm436
          Gene Name:
          predicted gene 436
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 144669937 - 144686368 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03057690_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm436
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085504.1 NP_001078973.1 2 - 3 388 79

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003950.mm.1

          Set Membership:

          Amplicon length between 71 and 85 3' Most Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          deacetylase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2133983 NM_001085504.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 43257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 172549 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477596 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233076 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 619091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 345774 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 183308 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152383 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 167958 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2133983 NM_001085504.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379403 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379409 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379388 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379360 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379412 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379380 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379363 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379394 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379383 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379358 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379425 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379422 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379396 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379369 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379417 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379366 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379391 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379419 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379378 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379429 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379432 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379405 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379419 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379409 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379410 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379430 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03589457 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03589453 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379422 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252597 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379425 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379424 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03666572 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379428 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379420 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252600 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252602 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379407 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379406 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379413 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03589454 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400032678.mm.1 TC0400003950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032678.mm.1 NM_001085504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032678.mm.1 NM_001085504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032679.mm.1 TC0400003950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032679.mm.1 NM_001085504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032679.mm.1 NM_001085504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032677.mm.1 TC0400003950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032677.mm.1 NM_001085504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032677.mm.1 NM_001085504.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032680.mm.1 TC0400003950.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032680.mm.1 NM_001085504.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032680.mm.1 NM_001085504.1 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_2529767 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline