Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03057780_m1
          See other GM13124 GE Assays ›
          Gene Symbol:
          Gm13124
          Gene Name:
          predicted gene 13124
          Gene Aliases:
          OTTMUSG00000010546
          Chromosome Location:
          Chr.4: 144555000 - 144565134 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03057780_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gm13124
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001085542.1 NP_001079011.1 2 - 3 380 61

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003949.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          deacetylase

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2148957 NM_001085542.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480648 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739379 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 537110 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 519650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 5235 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 370790 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 8230 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2148957 NM_001085542.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379057 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379073 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379030 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379023 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378997 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379025 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379083 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378960 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23378994 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379080 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23379006 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379088 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379078 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379083 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03450679 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379057 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379080 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379046 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379089 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379071 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03450678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379077 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03722186 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379050 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379036 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23378994 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23378991 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379059 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379043 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379048 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23378992 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379097 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379068 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379060 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03486503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03934107 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379052 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379082 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03772988 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379066 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379051 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03722185 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03450681 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03702592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23378993 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03450674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379041 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379073 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379091 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03957452 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379045 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04001370 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379092 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23379094 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03957456 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0400032675.mm.1 TC0400003949.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032675.mm.1 NM_001085542.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032675.mm.1 NM_001085542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032676.mm.1 TC0400003949.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032676.mm.1 NM_001085542.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032676.mm.1 NM_001085542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032673.mm.1 TC0400003949.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032673.mm.1 NM_001085542.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032673.mm.1 NM_001085542.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032674.mm.1 TC0400003949.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400032674.mm.1 NM_001085542.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400032674.mm.1 NM_001085542.1 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline