Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03057847_m1
          See other FAM159A GE Assays ›
          Gene Symbol:
          Fam159a
          Gene Name:
          family with sequence similarity 159, member A
          Gene Aliases:
          OTTMUSG00000008243
          Chromosome Location:
          Chr.4: 108367777 - 108383655 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03057847_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fam159a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001099303.2 NP_001092773.1 2 - 3 632 56

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003240.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 3' Most Recommended Assay Mus musculus Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2083499 NM_001099303.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 970365 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 216970 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56575 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 368105 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 945429 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 509512 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 728885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 56190 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 555973 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 16978 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152107 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2083499 NM_001099303.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195661 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195791 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195765 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195821 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195692 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195820 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195760 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195803 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195623 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195697 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195719 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195823 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195684 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195715 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195695 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195756 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195705 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195798 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195818 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195687 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195679 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195706 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195717 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195763 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195620 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195667 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195771 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195740 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195773 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195783 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195806 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195785 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195704 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23195632 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195617 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03807738 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195740 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03363311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03912277 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03353818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195710 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195767 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03562213 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299515 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195750 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03841137 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03907329 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195629 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195732 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195654 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195776 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195712 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04070488 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03324876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267585 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195803 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252111 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195624 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195785 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299532 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03901282 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03363301 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195749 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195751 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03782443 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03460809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195636 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195753 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03713093 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195705 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195637 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195692 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03457929 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03890703 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195744 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03765613 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195745 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195809 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03823953 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195725 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03901279 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195755 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195656 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299522 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195618 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195781 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195736 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04070491 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03252118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195706 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195649 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03353817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195651 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299533 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03907323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03584062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195688 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195689 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195643 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03363306 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195731 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03912274 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195621 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03807739 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03267586 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195713 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195793 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195792 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195623 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03979666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03299514 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195661 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195783 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195620 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195634 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23195721 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10515006 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515004 - -
          Affymetrix Mouse Gene 1.0 ST Array 10515005 - -
          Affymetrix Mouse Gene 1.1 ST Array 10515003 - -
          Affymetrix Mouse Clariom S Array PSR0400025810.mm.1 TC0400003240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400025810.mm.1 NM_001099303.2 Refseq

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline