Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03060106_gH
          See other MAGEA5 GE Assays ›
          Gene Symbol:
          Magea5
          Gene Name:
          melanoma antigen, family A, 5
          Gene Aliases:
          Mage-a5
          Chromosome Location:
          Chr.X: 155053061 - 155063145 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03060106_gH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Magea5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_020018.1 NP_064402.1 3 1503 99

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Magea5

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_66_P100794 NM_020018.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P100794 NM_020018.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P32758278 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P09465501 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P03780331 - -
          Whole Mouse Genome G4122A A_52_P120093 NM_020018.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P120093 BC109340.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P09465501 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P15043400 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P32758278 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P32758278 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10607456 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607453 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607445 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607451 - -
          Affymetrix Mouse Gene 1.0 ST Array 10607452 - -
          Affymetrix HT MG-430 PM Array Plate 1422325_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10607450 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607443 - -
          Affymetrix Mouse Gene 1.1 ST Array 10607454 - -
          Mouse Genome 430 2.0 Array 1422325_PM_at - -
          Mouse Genome 430 2.0 Array 1422325_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17141_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 17141_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0X00020225.mm.1 NM_020018.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020225.mm.1 NM_020018.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020225.mm.1 TC0X00003232.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020228.mm.1 NM_020018.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020228.mm.1 NM_020018.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020228.mm.1 TC0X00003232.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020224.mm.1 NM_020018.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020224.mm.1 NM_020018.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020224.mm.1 TC0X00003232.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0X00020226.mm.1 NM_020018.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020226.mm.1 NM_020018.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0X00020226.mm.1 TC0X00003232.mm.1 MTA TC
          Mouse Expression Set 430 1450542_PM_s_at - -
          Mouse Expression Set 430 1422325_PM_at - -
          Mouse Expression Set 430 1422325_at - -
          Mouse Expression Set 430 1450542_s_at - -
          Murine Genome U74v2 Set 103291_at - -
          Murine Genome U74v2 Set 103291_PM_at - -
          Mouse Genome 430A 2.0 Array 1422325_at - -
          Mouse Genome 430A 2.0 Array 1450542_s_at - -
          Mouse Genome 430A 2.0 Array 1422325_PM_at - -
          Mouse Genome 430A 2.0 Array 1450542_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1422325_at - -
          Affymetrix HT Mouse Genome 430A Array 1422325_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 901716 - -
          ABI Mouse Genome Survey Microarray 902627 - -
          ABI Mouse Genome Survey Microarray 903529 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 60193 - -
          MouseWG-6 v2 Expression BeadChip 6350039 - -
          Illumina mouse-6 v1.0 expression beadchip 6350039 - -
          Illumina mouse-6 v1.0 expression beadchip 60193 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0006350039 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000060193 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233373 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596441 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1233373 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596441 - -
          mouse-6 v1.1 (Illumina) scl0017141.1_282-S - -
          mouse-6 v1.1 (Illumina) scl017141.2_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 6350039 - -
          Illumina Sentrix mouse-6.v1 bead array 60193 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0017141.1_282-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl017141.2_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 6350039 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 60193 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596441 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2914608 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1233373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596441 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233373 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2914608 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline