Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03411735_sH
          See other OLFR456 GE Assays ›
          Gene Symbol:
          Olfr456
          Gene Name:
          olfactory receptor 456
          Gene Aliases:
          MOR257-7P
          Chromosome Location:
          Chr.6: 42486163 - 42487214 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03411735_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr456
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011528.2 NP_001011528.2 1 74 83
          GenBank mRNA BC139108.1 - 1 74 8862130 83
          BC139109.1 - 1 74 8862131 83

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0600002172.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr456

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476449 NM_001011528.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476449 BC139108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476449 BC139109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476449 NM_001011528.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476449 BC139108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476449 BC139109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476454 NM_001011528.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476454 BC139108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476454 BC139109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476454 NM_001011528.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476454 BC139108.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P476454 BC139109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476454 NM_001011528.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476454 BC139108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476454 BC139109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476454 NM_001011528.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476454 BC139108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476454 BC139109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476449 NM_001011528.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476449 BC139108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476449 BC139109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476449 NM_001011528.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476449 BC139108.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P476449 BC139109.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476454 NM_001011528.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476454 BC139108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476454 BC139109.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476454 NM_001011528.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476454 BC139108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476454 BC139109.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476449 NM_001011528.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476449 BC139108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476449 BC139109.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476449 NM_001011528.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476449 BC139108.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P476449 BC139109.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398659 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398634 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398613 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398632 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398614 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398638 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398644 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398611 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398668 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398620 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398630 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398664 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398642 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398654 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398651 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398649 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398666 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398612 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398637 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398660 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398640 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398629 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398646 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398662 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398636 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398617 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398658 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398652 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398639 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398643 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398656 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398619 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24398615 - -
          Whole Mouse Genome G4122A A_51_P476454 NM_001011528.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476454 BC139108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476454 BC139109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476454 NM_001011528.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476454 BC139108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P476454 BC139109.1 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05046343 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05412828 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05412841 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05046338 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05046352 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398612 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398598 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398636 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398594 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398643 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398651 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398653 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398600 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398587 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24398662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398662 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398620 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398611 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398636 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398613 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05104819 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398606 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05299831 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398660 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398649 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398594 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398628 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398630 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398629 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05323457 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398595 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05313915 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05412833 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05313913 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398639 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398651 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05046349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24398598 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10544439 - -
          Affymetrix Mouse Gene 1.0 ST Array 10544441 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544438 - -
          Affymetrix Mouse Gene 1.1 ST Array 10544440 - -
          Affymetrix Mouse Clariom S Array PSR0600017131.mm.1 TC0600002172.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017131.mm.1 NM_001011528.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017131.mm.1 NM_001011528.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017130.mm.1 TC0600002172.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0600017130.mm.1 NM_001011528.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0600017130.mm.1 NM_001011528.2 Refseq
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160851 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160851 - -
          mouse-6 v1.1 (Illumina) GI_58372127-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160851 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160851 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline