Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03414408_sH
          See other OLFR259 GE Assays ›
          Gene Symbol:
          Olfr259
          Gene Name:
          olfactory receptor 259
          Gene Aliases:
          MOR172-3
          Chromosome Location:
          Chr.2: 87107447 - 87108385 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03414408_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr259
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_146770.2 NP_666981.2 1 152 140

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0200003971.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr259

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          G-protein coupled receptor activity
          olfactory receptor activity
          odorant binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195598 NM_146770.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P195598 NM_146770.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140670 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102353 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195598 NM_146770.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P195598 NM_146770.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P195598 NM_146770.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P195598 NM_146770.2 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140670 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102353 - -
          Whole Mouse Genome G4122A A_51_P195598 NM_146770.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P195598 NM_146770.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103387 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102353 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140670 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404795 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404844 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404808 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404863 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404865 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404855 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404869 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404845 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404868 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404841 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404778 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404860 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404858 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404870 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404805 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404781 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404852 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404816 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404854 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404848 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404802 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404857 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404799 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404837 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404797 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404826 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404846 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21404824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404855 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404833 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404857 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404860 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404796 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404838 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404863 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404844 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404870 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404808 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404832 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02012383 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404837 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404812 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404795 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404802 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404865 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404846 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404852 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404835 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404797 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02080592 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404868 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404822 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02184747 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404839 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404824 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404875 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404848 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404827 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404858 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404826 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404854 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404834 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404798 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404845 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404874 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404799 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404869 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404864 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01629946 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21404815 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10484661 - -
          Affymetrix Mouse Gene 1.1 ST Array 10484660 - -
          Affymetrix Mouse Clariom S Array PSR0200033839.mm.1 NM_146770.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033839.mm.1 NM_146770.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200033839.mm.1 TC0200003971.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 832797 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5690746 - -
          Illumina mouse-6 v1.0 expression beadchip 5690746 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690746 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2596049 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2596049 - -
          mouse-6 v1.1 (Illumina) scl4318.1.1_330-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690746 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl4318.1.1_330-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690746 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2596049 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2596049 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline