Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03646986_g1
          See other HIST1H3F GE Assays ›
          Gene Symbol:
          Hist1h3f
          Gene Name:
          histone cluster 1, H3f
          Gene Aliases:
          H3.2-221, Hist1
          Chromosome Location:
          Chr.13: 23544052 - 23544954 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03646986_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Hist1h3f
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          GenBank mRNA BC101954.1 - 1 947 40044770 80
          BC101955.1 - 1 947 40044769 80
          BC101956.1 - 1 947 40044765 80
          BC103549.1 - 1 947 40044771 80

          Back To Top

          More Information


          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          histone

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882076 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027136 BC101955.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027136 BC101956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027136 BC103549.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027136 NM_013548.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2027136 BC101954.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027136 BC101955.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027136 BC101956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027136 BC103549.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027136 NM_013548.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2027136 BC101954.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28882076 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P105852 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10849893 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P10849894 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P28882076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882038 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882106 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882096 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882153 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882128 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882142 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882072 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882121 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28882110 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882097 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10560881 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882101 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882093 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10849893 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882087 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882092 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882096 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10849894 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P10560882 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882072 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882099 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P28882102 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10404066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10494406 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408203 - -
          Affymetrix Mouse Gene 1.0 ST Array 10404032 - -
          Affymetrix Mouse Gene 1.0 ST Array 10408240 - -
          Affymetrix HT MG-430 PM Array Plate 1431231_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10408239 - -
          Affymetrix Mouse Gene 1.1 ST Array 10408202 - -
          Affymetrix Mouse Gene 1.1 ST Array 10404030 - -
          Mouse Genome 430 2.0 Array 1431231_PM_at - -
          Mouse Genome 430 2.0 Array 1431231_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 260423_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 260423_at - -
          Mouse Expression Set 430 1427864_at - -
          Mouse Expression Set 430 1453573_PM_at - -
          Mouse Expression Set 430 1431231_PM_at - -
          Mouse Expression Set 430 1418366_at - -
          Mouse Expression Set 430 1431231_at - -
          Mouse Expression Set 430 1418367_x_at - -
          Mouse Expression Set 430 1418366_PM_at - -
          Mouse Expression Set 430 1418367_PM_x_at - -
          Mouse Expression Set 430 1460314_PM_s_at - -
          Mouse Expression Set 430 1453573_at - -
          Mouse Expression Set 430 1427864_PM_at - -
          Mouse Expression Set 430 1460314_s_at - -
          Mouse Expression Set 430 1442051_PM_at - -
          Mouse Expression Set 430 1442051_at - -
          Murine Genome U74v2 Set 93068_PM_r_at - -
          Murine Genome U74v2 Set 93023_PM_f_at - -
          Murine Genome U74v2 Set 93023_f_at - -
          Murine Genome U74v2 Set 93067_f_at - -
          Murine Genome U74v2 Set 94756_at - -
          Murine Genome U74v2 Set 94756_PM_at - -
          Murine Genome U74v2 Set 93022_i_at - -
          Murine Genome U74v2 Set 96416_f_at - -
          Murine Genome U74v2 Set 96416_PM_f_at - -
          Murine Genome U74v2 Set 93067_PM_f_at - -
          Murine Genome U74v2 Set 93022_PM_i_at - -
          Murine Genome U74v2 Set 93068_r_at - -
          Mouse Genome 430A 2.0 Array 1418366_PM_at - -
          Mouse Genome 430A 2.0 Array 1453573_PM_at - -
          Mouse Genome 430A 2.0 Array 1418366_at - -
          Mouse Genome 430A 2.0 Array 1453573_at - -
          Mouse Genome 430A 2.0 Array 1460314_s_at - -
          Mouse Genome 430A 2.0 Array 1431231_PM_at - -
          Mouse Genome 430A 2.0 Array 1460314_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1418367_x_at - -
          Mouse Genome 430A 2.0 Array 1427864_at - -
          Mouse Genome 430A 2.0 Array 1427864_PM_at - -
          Mouse Genome 430A 2.0 Array 1418367_PM_x_at - -
          Mouse Genome 430A 2.0 Array 1431231_at - -
          Affymetrix HT Mouse Genome 430A Array 1431231_at - -
          Affymetrix HT Mouse Genome 430A Array 1431231_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2320441 - -
          Illumina mouse-6 v1.0 expression beadchip 2480022 - -
          Illumina mouse-6 v1.0 expression beadchip 3940170 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002480022 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002320441 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003940170 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1213473 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2725359 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2703584 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2703584 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2725359 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1213473 - -
          mouse-6 v1.1 (Illumina) scl0260423.1_17-S - -
          mouse-6 v1.1 (Illumina) scl00260423.1_141-S - -
          mouse-6 v1.1 (Illumina) scl44997.1_85-S - -
          Illumina Sentrix mouse-6.v1 bead array 3940170 - -
          Illumina Sentrix mouse-6.v1 bead array 2480022 - -
          Illumina Sentrix mouse-6.v1 bead array 2320441 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0260423.1_17-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl00260423.1_141-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl44997.1_85-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2320441 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2480022 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3940170 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1213473 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2725359 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2925169 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1213473 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2725359 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2925169 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0