Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03647521_sH
          See other VMN2R103 GE Assays ›
          Gene Symbol:
          Vmn2r103
          Gene Name:
          vomeronasal 2, receptor 103
          Gene Aliases:
          EG627636, Vmn103
          Chromosome Location:
          Chr.17: 19773363 - 19812536 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03647521_sH
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Vmn2r103
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001104565.1 NP_001098035.1 6 2450 141

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Both primers and probe map within a single exon

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1962062 NM_001104565.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 480506 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 277292 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 190777 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 382877 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 156364 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 283815 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 23684 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 386044 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762810 XM_001472780.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 762810 XM_001480640.1 refseq_mrna_mouse_XM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 130729 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 32602 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 173272 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1962062 NM_001104565.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080563 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080575 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080560 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080628 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080634 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080723 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080555 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080607 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080534 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080711 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080677 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080685 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080664 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080713 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080542 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080573 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080561 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080520 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080604 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080517 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080614 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080516 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080595 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080540 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P31080550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080554 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080537 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13081646 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080561 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080593 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13185676 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13185677 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080575 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080565 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080582 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080573 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080534 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080560 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080555 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080578 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080583 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080550 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080528 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080532 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080588 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13074094 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080542 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080595 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080545 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P31080552 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P13244197 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10442111 - -
          Affymetrix Mouse Gene 1.1 ST Array 10442106 - -
          Affymetrix Mouse Clariom S Array PSR1700001844.mm.1 NM_001104565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001844.mm.1 NM_001104565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001844.mm.1 TC1700000240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001845.mm.1 NM_001104565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001845.mm.1 NM_001104565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001845.mm.1 TC1700000240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001840.mm.1 NM_001104565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001840.mm.1 NM_001104565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001840.mm.1 TC1700000240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001843.mm.1 NM_001104565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001843.mm.1 NM_001104565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001843.mm.1 TC1700000240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001841.mm.1 NM_001104565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001841.mm.1 NM_001104565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001841.mm.1 TC1700000240.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1700001842.mm.1 NM_001104565.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001842.mm.1 NM_001104565.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1700001842.mm.1 TC1700000240.mm.1 MTA TC
          Mouse Expression Set 430 1438124_PM_at - -
          Mouse Expression Set 430 1438124_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 753701 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1244091 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline