Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03647554_g1
          See other ANG4 GE Assays ›
          Gene Symbol:
          Ang4
          Gene Name:
          angiogenin, ribonuclease A family, member 4
          Gene Aliases:
          Raa4, Rnase5d
          Chromosome Location:
          Chr.14: 51763892 - 51773590 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03647554_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ang4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_177544.4 NP_808212.2 2 149 84
          GenBank mRNA AK136415.1 - 2 150 84
          AK165799.1 - 2 150 84
          AY219870.1 - 2 150 84
          BC042938.1 - 1 80 6398297 84
          BC132444.1 - 2 103 40130721 84
          BC132448.1 - 2 103 40130725 84

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ang4

          Gene Ontology Categories:

          Function(s) Process(es)

          angiogenesis
          RNA catabolic process
          positive regulation of cell proliferation
          antibacterial humoral response
          defense response to bacterium
          nucleic acid phosphodiester bond hydrolysis
          RNA phosphodiester bond hydrolysis
          nucleic acid binding
          nuclease activity
          endonuclease activity
          ribonuclease activity
          hydrolase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 NM_177544.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 AK136415.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 AK165799.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 AY219870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 BC042938.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 NM_177544.4 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 AK136415.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 AK165799.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 AY219870.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P98778 BC042938.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 739814 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 27693 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 NM_177544.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 AK136415.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 AK165799.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 AY219870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 BC042938.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 NM_177544.4 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 AK136415.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 AK165799.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 AY219870.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P98778 BC042938.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P502906 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603066 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P29603068 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P08111369 - -
          Whole Mouse Genome G4122A A_52_P98778 NM_177544.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 AK136415.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 AK165799.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 AY219870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 BC042938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 NM_177544.4 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 AK136415.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 AK165799.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 AY219870.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P98778 BC042938.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P502906 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132525 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P08111372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29602982 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29602979 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29602981 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29603068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P29602983 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603034 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603047 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603066 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603061 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603053 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603068 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603044 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P11137933 - -
          Agilent Six_ChIP_03_of_03_v2 A_68_P29603050 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10419577 - -
          Affymetrix Mouse Gene 1.0 ST Array 10419576 - -
          Affymetrix HT MG-430 PM Array Plate 1438364_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1440832_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10419575 - -
          Mouse Genome 430 2.0 Array 1440832_PM_at - -
          Mouse Genome 430 2.0 Array 1438364_PM_x_at - -
          Mouse Genome 430 2.0 Array 1440832_at - -
          Mouse Genome 430 2.0 Array 1438364_x_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 219033_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 219033_at - -
          Affymetrix Mouse Clariom S Array PSR1400015102.mm.1 NM_177544.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015102.mm.1 NM_177544.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015102.mm.1 TC1400002015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015101.mm.1 NM_177544.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015101.mm.1 NM_177544.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015101.mm.1 TC1400002015.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1400015103.mm.1 NM_177544.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015103.mm.1 NM_177544.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1400015103.mm.1 TC1400002015.mm.1 MTA TC
          Mouse Expression Set 430 1438364_PM_x_at - -
          Mouse Expression Set 430 1438364_x_at - -
          Mouse Expression Set 430 1440832_at - -
          Mouse Expression Set 430 1440832_PM_at - -
          Murine Genome U74v2 Set 168928_r_at - -
          Murine Genome U74v2 Set 168928_PM_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1440832_at - -
          Affymetrix HT Mouse Genome 430B Array 1438364_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1440832_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438364_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 705409 - -
          ABI Mouse Genome Survey Microarray 836575 - -
          Array Name Probe IDs Targets Target Type
          MouseRef-8 v2 Expression BeadChip ILMN_2819112 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2819112 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline