Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm03647560_s1
          See other FASTKD5 GE Assays ›
          Gene Symbol:
          Fastkd5
          Gene Name:
          FAST kinase domains 5
          Gene Aliases:
          C78212, mKIAA1792
          Chromosome Location:
          Chr.2: 130613838 - 130630027 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm03647560_s1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fastkd5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001146084.1 NP_001139556.1 3 2593 136
          NM_198176.2 NP_937819.2 3 2630 136
          GenBank mRNA AK143929.1 - 3 2569 136
          AK220420.1 - 1 2194 136
          BC048956.1 - 1 1632 136
          BC052898.1 - 3 2550 30059761 136

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fastkd5

          Gene Ontology Categories:

          Function(s) Process(es)

          cellular respiration
          protein kinase activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630827 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630829 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630828 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010161 BC048956.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010161 BC052898.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010161 AK143929.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010161 AW547061.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2010161 NM_001146084.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166129 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010161 BC048956.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010161 BC052898.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010161 AK143929.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010161 AW547061.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2010161 NM_001146084.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630828 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630827 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630830 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630826 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630821 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630829 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P21630824 - -
          Whole Mouse Genome G4122A A_52_P210933 BC052898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P210933 AK143929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P210933 AK220420.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P210933 NM_001146084.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P210933 NM_198176.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448866 BC048956.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448866 BC052898.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448866 AK143929.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448866 AK220420.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P448866 AW547061.2 mouse_transcriptome
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P01773119 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630828 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630829 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P21630827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630755 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630787 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630753 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630827 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630767 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630770 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630793 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630815 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630814 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630807 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630828 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630769 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630810 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630796 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630804 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630794 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630776 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630774 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630817 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P21630782 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01582648 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773121 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01415978 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01398669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01415980 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P02253673 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01673545 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630811 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630813 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773122 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773118 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630817 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630807 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01415979 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01965062 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01665391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630810 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01398671 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630814 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630806 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01773119 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630805 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630791 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01302398 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630804 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P01519756 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P21630815 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10487697 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487698 - -
          Affymetrix Mouse Gene 1.0 ST Array 10487699 - -
          Affymetrix HT MG-430 PM Array Plate 1435369_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10487690 - -
          Mouse Genome 430 2.0 Array 1435369_at - -
          Mouse Genome 430 2.0 Array 1435369_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380601_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 380601_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0200039612.mm.1 NM_001146084.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039612.mm.1 NM_001146084.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039612.mm.1 NM_198176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039612.mm.1 NM_198176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039612.mm.1 TC0200004631.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039615.mm.1 NM_198176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039615.mm.1 NM_198176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039615.mm.1 TC0200004631.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039611.mm.1 NM_001146084.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039611.mm.1 NM_001146084.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039611.mm.1 NM_198176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039611.mm.1 NM_198176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039611.mm.1 TC0200004631.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039614.mm.1 NM_001146084.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039614.mm.1 NM_001146084.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039614.mm.1 NM_198176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039614.mm.1 NM_198176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039614.mm.1 TC0200004631.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0200039613.mm.1 NM_001146084.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039613.mm.1 NM_001146084.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039613.mm.1 NM_198176.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039613.mm.1 NM_198176.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0200039613.mm.1 TC0200004631.mm.1 MTA TC
          Mouse Expression Set 430 1435369_PM_at - -
          Mouse Expression Set 430 1435369_at - -
          Murine Genome U74v2 Set 104157_at - -
          Murine Genome U74v2 Set 104157_PM_at - -
          Mouse Genome 430A 2.0 Array 1435369_at - -
          Mouse Genome 430A 2.0 Array 1435369_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1435369_at - -
          Affymetrix HT Mouse Genome 430A Array 1435369_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 1500162 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001500162 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2716247 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2716247 - -
          mouse-6 v1.1 (Illumina) scl0380601.1_239-S - -
          Illumina Sentrix mouse-6.v1 bead array 1500162 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0380601.1_239-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1500162 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2716247 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2716247 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0380601.1_239-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0